VWR Collection Laboratory Equipment - page 56
54
Description
Electrical (50/60 Hz)
Positions
Cat. No.
Standard High Volume Stirrer
120V | 1.3 amps | 75 watts
1
Standard High Volume Stirrer
230V | 0.65 amps | 75 watts
1
VWR
®
Standard High Volume Stirrer
Description
Electrical (50/60 Hz)
Positions
Cat. No.
Advanced High Volume Stirrer
120V | 1.3 amps | 75 watts
1
Advanced High Volume Stirrer
230V | 0.65 amps | 75 watts
1
VWR
®
Advanced High Volume Stirrer
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). Units are also supplied with a 2.75” (7cm) PTFE
coated stir bar. 2 year limited warranty on parts and labor.
• Powerful motor and magnet mix up to 25L of liquid
• Large top plate surface
• Microprocessor controls
VWR
®
Standard and Advanced High Volume Stirrers are designed for large
capacity applications. Powerful magnet and motor offer exceptional magnetic
coupling force capable of stirring 25L.
Operating Features:
Low profile design:
Takes up minimal space. Spill-resistant housing channels
fluids away from internal components. Cast aluminum base offers durability and
added stability.
Glass filled nylon top plate:
Features a chemical resistant, reflective white top
plate surface that is easy to clean.
Large top plate:
Accommodates oversized vessels.
Microprocessor control:
Precisely regulates speed through the entire range.
Stirring function, with continuous duty motor and magnet, maintains set speed even
under changing load or viscosity.
Adjustment knob:
Basic speed control knob with dial markings from 1 to 10
(Standard Series).
LED display:
Touch pad controls with easy-to-read, independent LED displays
for speed and time. Speed is adjustable in 10rpm increments. Timer will display
elapsed time or, when programmed to user defined limit, will shut off unit when time
reaches zero (Advanced Series).
Safety Features:
Audible alarm:
In timed mode, alarm will sound when time reaches zero.
(Advanced Series)
Speed ramping feature:
Slowly increases speed for improved safety and enhanced
coupling. Avoids splashing, improves spin bar control and provides excellent low
speed control.
Operating Conditions:
Units can be run in conditions from 5 to 40°C (41 to 104°F), maximum 80% relative
humidity, non-condensing.
Applications:
Carboys, chromatography, and large volume sample stirring.
VWR
®
Standard & Advanced High Volume Stirrers
VWR
®
HOTPLATES/STIRRERS
Specifications
Speed Range
60 to 1400rpm
Speed Stability
± 2%
Timer (Advanced Series)
1 second to 160 hours
Maximum Capacity (H
2
O)
25L
Maximum Vessel Diameter
10” (25cm)
Top Plate Material
Glass filled nylon
Top Plate Dimensions (L x W)
12.5 x 11” (31.8 x 27.9cm)
Overall Dimensions (L x W x H)
17 x 11 x 4”
(43.2 x 27.9 x 10.2cm)
Ship Weight
14.6lbs (6.6kg)
1...,46,47,48,49,50,51,52,53,54,55 57,58,59,60,61,62,63,64,65,66,...116
Powered by FlippingBook