VWR Collection Laboratory Equipment - page 55
53
• Hotplates & Hotplate-Stirrers boil 300mL of water in 12 minutes
• Ideal for educational labs
• Built in support rod holder
VWR
®
Basic Mini Hotplates, Stirrers, and Hotplate-Stirrers are rugged, compact
units that heat and stir up to 1000mL of liquid. Durable, cast aluminum top plate
will not crack or chip, and provides an even heating surface. Bi-metallic thermostat
offers reliable temperature control. Powerful heater reaches maximum temperature
in only minutes. Powerful motor and magnet deliver reliable and consistent stirring.
Compact design saves bench space. Built in support rod holder with locking knob
accepts optional Support Rod and Clamp Kit.
Basic Mini Fixed Temperature Hotplate features an illuminated rocker switch to
activate the preset fixed temperature of 400°C.
Basic Mini Auto-Stirrer is automatically activated by the minimum weight of ~50
grams (1.75oz) and will stop stirring when mixing vessel is removed.
Operating Features:
Adjustment knobs:
Basic speed and temperature control knob with dial markings
from 1 to 10.
Operating Conditions:
Units can be run in conditions from 5 to 40°C (41 to 104°F), maximum 85% relative
humidity, non-condensing.
Applications:
Academia, Chemistry 101
Ordering Information
Ordering Information Units include a 72” (183cm), 3-wire cord and plug (230 volt
units are supplied with Euro type plug). Stirrers and Hotplate-Stirrers are supplied
with a 1.5” (3.8cm) PTFE coated stir bar. 2 year limited warranty on parts and labor.
Visit
for additional colors.
Description
Electrical (50/60 Hz)
Cat. No.
Basic Mini Hotplate
120V | 6 amps | 565 watts
Basic Mini Hotplate
230V | 2.5 amps | 430 watts
Basic Mini Fixed Temperature Hotplate
120V | 6 amps | 565 watts
VWR
®
Basic Mini Hotplates
Description
Electrical (50/60 Hz)
Cat. No.
Basic Mini Stirrer
120V | 0.5 amps | 20 watts
Basic Mini Stirrer
230V | 0.25 amps | 20 watts
Basic Mini Auto-Stirrer
120V | 0.5 amps | 20 watts
Basic Mini Auto-Stirrer
230V | 0.25 amps | 20 watts
VWR
®
Basic Mini Stirrers
Description
Electrical (50/60 Hz)
Cat. No.
Basic Mini Hotplate-Stirrer
120V | 6 amps | 565 watts
Basic Mini Hotplate-Stirrer
230V | 2.5 amps | 430 watts
VWR
®
Basic Mini Hotplate-Stirrers
* Fixed Temperature Hotplate has a fixed temperature of 400°C.
VWR
®
Basic Mini Hotplates, Stirrers & Hotplate-Stirrers
VWR
®
HOTPLATES/STIRRERS
Specifications
Temperature Range
to 400°C *
Speed Range
100 to 1200rpm
Maximum Capacity (H
2
O)
1000mL
Top Plate Dimensions (Dia.)
4.5” (11.4cm)
Overall Dimensions (L x W x H)
6.5 x 5.75 x 5.4”
(16.5 x 14.6 x 13.7cm)
Ship Weight
4.4lbs (2kg)
Stirrer
Hotplate-Stirrer
Hotplate
1...,45,46,47,48,49,50,51,52,53,54 56,57,58,59,60,61,62,63,64,65,...116
Powered by FlippingBook