VWR Collection Laboratory Equipment - page 54
52
UNI-BLOCKS
Test Tubes
Sample Type
No. of Wells
Well Dia.
Well Depth
Cat. No.
12mm Tube
40
12.7mm
45.7mm
16mm Tube
32
17.5mm
45.7mm
20mm Tube
32
20.5mm
45.7mm
25mm Tube
24
25.4mm
41.9mm
• Heat and stir a variety of samples simultaneously on one hotplate
• Uni and sectional blocks accommodate 9 different sample sizes
• Anodized aluminum with close block to tube contact ensures optimal transfer of heat
The VWR
®
Multi-Sample Reaction Station enables you to heat and stir multiple samples of one size or up to
five multiple samples of different sizes, all with one unit. The base plate can hold 5 sectional blocks or one
uni-block. For added versatility, the base plate has a centrally located threaded opening to accommodate a
1/2” (13mm) support rod. Both block styles are offered in 9 different sample size configurations for vials and
test tubes. Blocks are made of anodized aluminum to provide superior temperature stability and heat transfer.
Each block has a thermometer well for measuring block temperature. Base plate is designed to fit on the VWR
®
Professional or Advanced Round Top or other 135mm diameter top models. Safety handles are available to
aid in the safe removal of the blocks or base plate.
Sample Type
No. of Wells
Well Dia.
Well Depth
Cat. No.
12mm Vial (2mL)
40
12.7mm
16.8mm
15mm Vial (1 dram)
40
15.5mm
16.8mm
17mm Vial (2 dram)
32
17.8mm
16.8mm
21mm Vial (4 dram)
24
21.5mm
16.8mm
28mm Vial
16
28.8mm
16.8mm
Vials
SECTIONAL BLOCKS
Test Tubes
Sample Type
No. of Wells
Well Dia.
Well Depth
Cat. No.
12mm Tube
9
12.7mm
45.7mm
16mm Tube
8
17.5mm
45.7mm
20mm Tube
5
20.5mm
45.7mm
25mm Tube
5
25.4mm
41.9mm
Sample Type
No. of Wells
Well Dia.
Well Depth
Cat. No.
12mm Vial (2mL)
9
12.7mm
16.8mm
15mm Vial (1 dram)
10
15.5mm
16.8mm
17mm Vial (2 dram)
10
17.8mm
16.8mm
21mm Vial (4 dram)
5
21.5mm
16.8mm
28mm Vial
3
28.8mm
16.8mm
Vials
Other Accessories
Accessory
Cat. No.
Base plate for 135mm diameter plates
Safety Handles (2)
VWR
®
Round Top Hotplate-Stirrer Accessories
VWR
®
HOTPLATES/STIRRERS
1...,44,45,46,47,48,49,50,51,52,53 55,56,57,58,59,60,61,62,63,64,...116
Powered by FlippingBook