VWR Collection Laboratory Equipment - page 49
47
VWR
®
Advanced Hotplates & Hotplate-Stirrers
VWR
®
HOTPLATES/STIRRERS
VWR
®
Advanced Hotplates
Description
Top Plate Material
Electrical (50/60 Hz)
Cat. No.
4 x 4” (10.2 x 10.2cm) Hotplate
Ceramic
120V | 2.9 amps | 350 watts
4 x 4” (10.2 x 10.2cm) Hotplate
Ceramic
230V | 1.5 amps | 350 watts
4 x 4” (10.2 x 10.2cm) Hotplate
Aluminum
120V | 2.9 amps | 350 watts
4 x 4” (10.2 x 10.2cm) Hotplate
Aluminum
230V | 1.5 amps | 350 watts
7 x 7” (17.8 x 17.8cm) Hotplate
Ceramic
120V | 7.9 amps | 950 watts
7 x 7” (17.8 x 17.8cm) Hotplate
Ceramic
230V | 4.4 amps | 1000 watts
7 x 7” (17.8 x 17.8cm) Hotplate
Aluminum
120V | 7.9 amps | 950 watts
7 x 7” (17.8 x 17.8cm) Hotplate
Aluminum
230V | 4.4 amps | 1000 watts
10 x 10” (25.4 x 25.4cm) Hotplate
Ceramic
120V | 12.5 amps | 1500 watts
10 x 10” (25.4 x 25.4cm) Hotplate
Ceramic
230V | 6.7 amps | 1550 watts
10 x 10” (25.4 x 25.4cm) Hotplate
Aluminum
120V | 12.5 amps | 1500 watts
10 x 10” (25.4 x 25.4cm) Hotplate
Aluminum
230V | 6.7 amps | 1550 watts
VWR
®
Advanced Hotplate-Stirrers
Description
Top Plate Material
Electrical (50/60 Hz)
Cat. No.
4 x 4” (10.2 x 10.2cm) Hotplate-Stirrer
Ceramic
120V | 3.3 amps | 400 watts
4 x 4” (10.2 x 10.2cm) Hotplate-Stirrer
Ceramic
230V | 1.7 amps | 400 watts
4 x 4” (10.2 x 10.2cm) Hotplate-Stirrer
Aluminum
120V | 3.3 amps | 400 watts
4 x 4” (10.2 x 10.2cm) Hotplate-Stirrer
Aluminum
230V | 1.7 amps | 400 watts
7 x 7” (17.8 x 17.8cm) Hotplate-Stirrer
Ceramic
120V | 8.3 amps | 1000 watts
7 x 7” (17.8 x 17.8cm) Hotplate-Stirrer
Ceramic
230V | 4.6 amps | 1050 watts
7 x 7” (17.8 x 17.8cm) Hotplate-Stirrer
Aluminum
120V | 8.3 amps | 1000 watts
7 x 7” (17.8 x 17.8cm) Hotplate-Stirrer
Aluminum
230V | 4.6 amps | 1050 watts
10 x 10” (25.4 x 25.4cm) Hotplate-Stirrer
Ceramic
120V | 12.9 amps | 1550 watts
10 x 10” (25.4 x 25.4cm) Hotplate-Stirrer
Ceramic
230V | 7.0 amps | 1600 watts
10 x 10” (25.4 x 25.4cm) Hotplate-Stirrer
Aluminum
120V | 12.9 amps | 1550 watts
10 x 10” (25.4 x 25.4cm) Hotplate-Stirrer
Aluminum
230V | 7.0 amps | 1600 watts
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). Stirrers and Hotplate-Stirrers are supplied with a 1.5”
(3.8cm) PTFE coated stir bar. 2 year limited warranty on parts and labor.
Optional Support Rod and Clamp Kit:
Kit includes 18” (45.7cm) stainless steel support rod, 3-prong medium swivel clamp, thermometer/temperature probe extension
clamp, and hook connector (see page 61).
1...,39,40,41,42,43,44,45,46,47,48 50,51,52,53,54,55,56,57,58,59,...116
Powered by FlippingBook