VWR Collection Laboratory Equipment - page 47
45
VWR
®
Professional Hotplates, Stirrers & Hotplate-Stirrers
VWR
®
HOTPLATES/STIRRERS
VWR
®
Professional Hotplates
Description
Top Plate Material
Electrical (50/60 Hz)
Cat. No.
7 x 7” (17.8 x 17.8cm) Hotplate
Ceramic
120V | 7.9 amps | 950 watts
7 x 7” (17.8 x 17.8cm) Hotplate
Ceramic
230V | 4.4 amps | 1000 watts
7 x 7” (17.8 x 17.8cm) Hotplate
Aluminum
120V | 7.9 amps | 950 watts
7 x 7” (17.8 x 17.8cm) Hotplate
Aluminum
230V | 4.4 amps | 1000 watts
7 x 7” (17.8 x 17.8cm) Hotplate with NIST Traceable Certificate
Ceramic
120V | 7.9 amps | 950 watts
7 x 7” (17.8 x 17.8cm) Hotplate with NIST Traceable Certificate
Ceramic
230V | 4.4 amps | 1000 watts
7 x 7” (17.8 x 17.8cm) Hotplate with NIST Traceable Certificate
Aluminum
120V | 7.9 amps | 950 watts
7 x 7” (17.8 x 17.8cm) Hotplate with NIST Traceable Certificate
Aluminum
230V | 4.4 amps | 1000 watts
10 x 10” (25.4 x 25.4cm) Hotplate
Ceramic
120V | 12.5 amps | 1500 watts
10 x 10” (25.4 x 25.4cm) Hotplate
Ceramic
230V | 6.7 amps | 1550 watts
10 x 10” (25.4 x 25.4cm) Hotplate
Aluminum
120V | 12.5 amps | 1500 watts
10 x 10” (25.4 x 25.4cm) Hotplate
Aluminum
230V | 6.7 amps | 1550 watts
10 x 10” (25.4 x 25.4cm) Hotplate with NIST Traceable Certificate
Ceramic
120V | 12.5 amps | 1500 watts
10 x 10” (25.4 x 25.4cm) Hotplate with NIST Traceable Certificate
Ceramic
230V | 6.7 amps | 1550 watts
10 x 10” (25.4 x 25.4cm) Hotplate with NIST Traceable Certificate
Aluminum
120V | 12.5 amps | 1500 watts
10 x 10” (25.4 x 25.4cm) Hotplate with NIST Traceable Certificate
Aluminum
230V | 6.7 amps | 1550 watts
VWR
®
Professional Stirrers
Description
Top Plate Material
Electrical (50/60 Hz)
Cat. No.
7 x 7” (17.8 x 17.8cm) Stirrer
Ceramic
120V | 0.4 amps | 50 watts
7 x 7” (17.8 x 17.8cm) Stirrer
Ceramic
230V | 0.2 amps | 50 watts
7 x 7” (17.8 x 17.8cm) Stirrer
Aluminum
120V | 0.4 amps | 50 watts
7 x 7” (17.8 x 17.8cm) Stirrer
Aluminum
230V | 0.2 amps | 50 watts
10 x 10” (25.4 x 25.4cm) Stirrer
Ceramic
120V | 0.4 amps | 50 watts
10 x 10” (25.4 x 25.4cm) Stirrer
Ceramic
230V | 0.2 amps | 50 watts
10 x 10” (25.4 x 25.4cm) Stirrer
Aluminum
120V | 0.4 amps | 50 watts
10 x 10” (25.4 x 25.4cm) Stirrer
Aluminum
230V | 0.2 amps | 50 watts
VWR
®
Professional Hotplate-Stirrers
Description
Top Plate Material
Electrical (50/60 Hz)
Cat. No.
7 x 7” (17.8 x 17.8cm) Hotplate-Stirrer
Ceramic
120V | 8.3 amps | 1000 watts
7 x 7” (17.8 x 17.8cm) Hotplate-Stirrer
Ceramic
230V | 4.6 amps | 1050 watts
7 x 7” (17.8 x 17.8cm) Hotplate-Stirrer
Aluminum
120V | 8.3 amps | 1000 watts
7 x 7” (17.8 x 17.8cm) Hotplate-Stirrer
Aluminum
230V | 4.6 amps | 1050 watts
7 x 7” (17.8 x 17.8cm) Hotplate-Stirrer with NIST Traceable Certificate
Ceramic
120V | 8.3 amps | 1000 watts
7 x 7” (17.8 x 17.8cm) Hotplate-Stirrer with NIST Traceable Certificate
Ceramic
230V | 4.6 amps | 1050 watts
7 x 7” (17.8 x 17.8cm) Hotplate-Stirrer with NIST Traceable Certificate
Aluminum
120V | 8.3 amps | 1000 watts
7 x 7” (17.8 x 17.8cm) Hotplate-Stirrer with NIST Traceable Certificate
Aluminum
230V | 4.6 amps | 1050 watts
10 x 10” (25.4 x 25.4cm) Hotplate-Stirrer
Ceramic
120V | 12.9 amps | 1550 watts
10 x 10” (25.4 x 25.4cm) Hotplate-Stirrer
Ceramic
230V | 7.0 amps | 1600 watts
10 x 10” (25.4 x 25.4cm) Hotplate-Stirrer
Aluminum
120V | 12.9 amps | 1550 watts
10 x 10” (25.4 x 25.4cm) Hotplate-Stirrer
Aluminum
230V | 7.0 amps | 1600 watts
10 x 10” (25.4 x 25.4cm) Hotplate-Stirrer with NIST Traceable Certificate
Ceramic
120V | 12.9 amps | 1550 watts
10 x 10” (25.4 x 25.4cm) Hotplate-Stirrer with NIST Traceable Certificate
Ceramic
230V | 7.0 amps | 1600 watts
10 x 10” (25.4 x 25.4cm) Hotplate-Stirrer with NIST Traceable Certificate
Aluminum
120V | 12.9 amps | 1550 watts
10 x 10” (25.4 x 25.4cm) Hotplate-Stirrer with NIST Traceable Certificate
Aluminum
230V | 7.0 amps | 1600 watts
Optional NIST traceable certificate:
Includes a 3 point NIST traceable calibration. The traceable certificate includes actual calibration measurement data and uncertainty.
The calibration laboratory is ISO/IEC 17025 compliant (for hotplates and hotplate-stirrers).
Replacement probes
are available, see accessories on page 59.
1...,37,38,39,40,41,42,43,44,45,46 48,49,50,51,52,53,54,55,56,57,...116
Powered by FlippingBook