VWR Collection Laboratory Equipment - page 43
41
Test Tube Racks - Full Size, Stationary
• PVC coated steel
• Includes hardware for easy attachment
to platforms
• 3.75L x 16.5W x 4H” (9.5 x 41.9 x 10.2cm)
Test Tube Racks - Full Size, Pivoting
• Adjustable angle, custom tilt
• Stainless steel holder includes removable rack
• Rack is easily removed to transport from work
area to shaker
• Dimensions (L x W x H)
Inside Rack: 5.1 x 10.8 x 3.9” (12.9 x 27.4 x 9.9cm)
Outer Rack: 5 x 10.9 x 5” (12.7 x 27.6 x 12.7cm)
VWR
®
SHAKERS
VWR
®
Test Tube Racks
10 to 14mm
Test Tube Rack
Full Size, Stationary
16 to 20mm
Test Tube Rack
Full Size, Stationary
21 to 25mm
Test Tube Rack
Full Size, Stationary
50mL Centrifuge
Tube Rack
Full Size, Stationary
13mm
Test Tube Rack
Full Size, Pivoting
16mm
Test Tube Rack
Full Size, Pivoting
20mm
Test Tube Rack
Full Size, Pivoting
25mm
Tube Rack
Full Size, Pivoting
30mm
Tube Rack
Full Size, Pivoting
Tube Capacity = 48 Tube Capacity = 33 Tube Capacity = 21 Tube Capacity = 17 Tube Capacity = 90 Tube Capacity = 60 Tube Capacity = 40 Tube Capacity = 24 Tube Capacity = 21
Number of Test Tube Racks per Tray or Platform
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
N/A
1
1
1
1
1
N/A
N/A
N/A
N/A
1
1
1
1
1
N/A
N/A
N/A
N/A
1
1
1
1
1
N/A
N/A
N/A
N/A
2
2
2
2
2
N/A
N/A
N/A
N/A
2
2
2
2
2
3
3
3
3
2
2
2
2
2
3
3
3
3
2
2
2
2
2
3
3
3
3
2
2
2
2
2
5
5
5
5
3
3
3
3
3
5
5
5
5
3
3
3
3
3
5
5
5
5
3
3
3
3
3
6
6
6
6
4
4
4
4
4
7
7
7
7
4
4
4
4
4
5
5
5
5
6
6
6
6
6
1...,33,34,35,36,37,38,39,40,41,42 44,45,46,47,48,49,50,51,52,53,...116
Powered by FlippingBook