VWR Collection Laboratory Equipment - page 42
40
Test Tube Racks - Half Size, Stationary
• PVC coated steel
• Includes hardware for easy attachment to platforms
• 5L x 7W x 4H” (12.7 x 17.8 x 10.2cm) / Micro-Tube Rack: 1.7H” (4.3cm)
VWR
®
SHAKERS
VWR
®
Test Tube Racks
1.5 to 2mL
Micro-Tube Rack
Half Size, Stationary
10 to 13mm
Test Tube Rack
Half Size, Stationary
14 to 16mm
Test Tube Rack
Half Size, Stationary
18 to 20mm
Test Tube Rack
Half Size, Stationary
22 to 25mm
Test Tube Rack
Half Size, Stationary
15mL Centrifuge
Tube Rack
Half Size, Stationary
50mL Centrifuge
Tube Rack
Half Size, Stationary
Tube Capacity = 70 Tube Capacity = 63 Tube Capacity = 48 Tube Capacity = 35 Tube Capacity = 24 Tube Capacity = 35 Tube Capacity = 12
Tray or Platform
Number of Test Tube Racks per Tray or Platform
11.75 x 8.75” (29.9 x 22.2cm)
Mini Shaker, Mini Shaker 15 *
2
2
2
2
2
2
2
11 x 7.75” (27.9 x 19.7cm)
Incubating Mini Shaker *
2
1
1
N/A
N/A
1
1
Microplate Shaker *
2
N/A
N/A
N/A
N/A
N/A
N/A
Incubating Microplate Shaker *
2
N/A
N/A
N/A
N/A
N/A
N/A
11 x 13” (27.9 x 33cm)
Standard/Advanced 3500
2
2
2
2
2
2
2
Professional 3500
2
2
2
2
2
2
2
Advanced 3750
2
2
2
2
2
2
2
13 x 13” (33cm x 33cm)
Standard/Advanced 3500
2
2
2
2
2
2
2
Advanced 3750
2
2
2
2
2
2
2
18 x 18” (45.7 x 45.7cm)
Standard/Advanced 3500
4
4
4
4
4
4
4
Advanced 3750
4
4
4
4
4
4
4
5000I / 5000IR
4
4
4
4
4
4
4
18 x 24” (45.7 x 61cm)
Standard/Advanced 3500
6
6
6
6
6
6
6
Advanced 3750
6
6
6
6
6
6
6
Standard/Advanced 5000
6
6
6
6
6
6
6
18 x 30” (45.7 x 76.2cm)
Standard/Advanced 5000
8
8
8
8
8
8
8
24 x 24” (61 x 61cm)
Advanced 10000-1/10000-2
8
8
8
8
8
8
8
24 x 36” (61 x 91.4cm)
Advanced 15000-1/15000-2
7
7
7
7
7
7
7
* All units require an accessory/platform for mounting flask clamps or test tube racks with the exception of the Mini Shaker, Mini Shaker 15, Microplate Shaker, Incubating Mini Shaker and Incubating
Microplate Shaker
1...,32,33,34,35,36,37,38,39,40,41 43,44,45,46,47,48,49,50,51,52,...116
Powered by FlippingBook