VWR Collection Laboratory Equipment - page 40
38
Stainless Steel Flask Clamps
• Designed to hold Erlenmeyer flasks from 10mL to 6L
• Constructed of type 302 and 304 stainless steel
• Includes hardware for easy attachment to platforms
• Flask clamps 125mL and higher are supplied with a spring to hold
flask in place
• 2.8L clamp is designed to hold a Fernbach flask
• Media bottle clamps feature the same details as flask clamps
Erlenmeyer Flask Clamps
Media Bottle Flask Clamps
VWR
®
SHAKERS
VWR
®
Flask Clamps
10mL
Erlenmeyer
Flask Clamp
25mL
Erlenmeyer
Flask Clamp
50mL
Erlenmeyer
Flask Clamp
125mL
Erlenmeyer
Flask Clamp
250mL
Erlenmeyer
Flask Clamp
500mL
Erlenmeyer
Flask Clamp
1L
Erlenmeyer
Flask Clamp
2L
Erlenmeyer
Flask Clamp
Stainless Steel
Stainless Steel
Stainless Steel
Stainless Steel
Stainless Steel
Stainless Steel
Stainless Steel
Stainless Steel
Tray or Platform
Number of Flask Clamps per Tray or Platform
11.75 x 8.75” (29.9 x 22.2cm)
Mini Shaker, Mini Shaker 15 *
35
20
15
12
6
4
N/A
N/A
11 x 7.75” (27.9 x 19.7cm)
Incubating Mini Shaker *
35
20
12
8
5
N/A
N/A
N/A
11 x 13” (27.9 x 33cm)
Standard/Advanced 3500
60
25
13
10
9
7
4
N/A
Professional 3500
60
25
13
10
9
7
4
N/A
Advanced 3750
60
25
13
10
9
7
4
N/A
13 x 13” (33cm x 33cm)
Standard/Advanced 3500
60
30
15
12
12
8
4
3
Advanced 3750
60
30
15
12
12
8
4
3
18 x 18” (45.7 x 45.7cm)
Standard/Advanced 3500
113
64
32
20
20
13
8
5
Advanced 3750
113
64
32
20
20
13
8
5
5000I / 5000IR
113
64
32
20
20
13
8
5
18 x 24” (45.7 x 61cm)
Standard/Advanced 3500
158
88
44
28
28
20
12
6
Advanced 3750
158
88
44
28
28
20
12
6
Standard/Advanced 5000
158
88
44
28
28
20
12
6
18 x 30” (45.7 x 76.2cm)
Standard/Advanced 5000
203
112
56
36
36
26
15
8
24 x 24” (61 x 61cm)
Advanced 10000-1/10000-2
221
121
61
41
41
25
16
9
24 x 36” (61 x 91.4cm)
Advanced 15000-1/15000-2
336
160
94
61
64
40
24
14
* All units require an accessory/platform for mounting flask clamps or test tube racks with the exception of the Mini Shaker, Mini Shaker 15 and Incubating Mini Shaker
1...,30,31,32,33,34,35,36,37,38,39 41,42,43,44,45,46,47,48,49,50,...116
Powered by FlippingBook