VWR Collection Laboratory Equipment - page 4
2
The new VWR
®
family of laboratory shakers has one of the industries largest offerings on the market today, covering a wide range of applications. Many shakers
are available as incubating, cooling, or open air and with orbital, reciprocating, rocking, or waving motions. Capacities range from one microplate up to 150lbs
(68.1kg).
• VWR
®
introduces
ALL NEW TOUCH SCREEN TECHNOLOGY
2 new models; the VWR
®
Thermal Shake Touch and the VWR
®
Cooling Thermal Shake Touch...operating a shaker this
sophisticated has never been easier.
• Touch screen technology
• On board help screens available on almost every screen
• Programmable for up to 5 separate programs or 10 if using the USB
• Small footprint
• Rapid heating and cooling with high speed shaking
• Patented design features
• Exceptional extended warranty on most models
• Advanced electronics
• Safety features
• Calibration modes
• Interfacing
• Easy to use controls
• Expanded line of accessories to fit almost any application
• Platforms (available in a variety of sizes):
• Universal - lets you easily mount flask clamps and test tube racks
• Dedicated - pre-mounted flask clamps of the same size, maximizes the capacity of your platform
• Adjustable - uses roller bars allowing the most flexibility in securing your vessels
• Culture- ideal for slow speed applications utilizing low profile, flat bottom vessels
• Large Vessel Carrier - utilizes high sides and a heavy-duty design to help secure oversized vessels
• Separatory Funnel - holds three 500mL to 2L funnels
• Clamps
• Erlenmeyer
• Media bottles
• Microplates
• Test Tube Racks
• Pivoting
• Fixed position
VWR
®
SHAKERS
1,2,3 5,6,7,8,9,10,11,12,13,14,...116
Powered by FlippingBook