VWR Collection Laboratory Equipment - page 39
37
Universal Harness
• Attaches to tray to secure low profile plates
Description
Used on Unit
Cat. No.
Stacking Tray, 12.75 x 10”
Rocking Platform Shaker
(32.4 x 25.4cm)
Stacking Tray, 11.75 x 8.75”
3-D Rotator Waver
(29.9 x 22.2cm)
Stacking Tray
Easily attaches to the units included tray to add a second tier for higher capacity applications.
Second tier tray mounts 3.5” (8.9cm) above lower tray and includes a non-skid rubber mat.
Description
Used on Shakers
Cat. No.
Universal Harness
Mini Shaker, Mini Shaker 15
Dimpled Mat
Designed to hold centrifuge tubes, vials, culture tubes, and micro-tubes securely in place. Mat can
be easily removed for cleaning and transporting of tubes from bench to tray.
Description
Used on Unit Cat. No.
Dimpled Mat, 12.75 x 10”
Rocking Platform Shaker
(32.4 x 25.4cm)
Dimpled Mat, 11.75 x 8.75”
3-D Rotator Waver
(29.9 x 22.2cm)
Dimpled Mat, 10 x 7.5”
Incubating Rocking Platform Shaker
(25.4 x 19.1cm)
Dimpled Mat, 9.25 x 7.25”
Incubating 3-D Rotator Waver
(23.5 x 18.4cm)
Dilution Cup Tray
• Constructed of type 304 stainless steel
• Holds 24 x 28mm dilution vials
Description
Used on Unit
Cat. No.
Dilution Rack Tray, 6.9 x 10”
Mini Shaker, Mini Shaker 15
(17.5 x 25.4cm)
VWR
®
SHAKERS
VWR
®
Accessories
Description
Used on Unit
Cat. No.
1.5 to 2mL Micro-Tube Rack
Mini Shaker, Mini Shaker 15,
Microplate Shaker, Incubating Microplate Shaker
Micro-Tube Rack
Optional 1.5 to 2mL Micro-Tube Rack attaches to tray to hold up to 70, 1.5mL or 2mL micro-tubes.
Tray can accommodate up to 2 micro-tube racks.
1...,29,30,31,32,33,34,35,36,37,38 40,41,42,43,44,45,46,47,48,49,...116
Powered by FlippingBook