VWR Collection Laboratory Equipment - page 38
36
Description
Bar Size
Overall Dimensions (L x W x H)
Used on Shakers
Cat. No.
4-bar Large Vessel Carrier Platform 18” (45.7cm)
30.1 x 18.1 x 13.9 (76.4 x 45.9 x 93.7cm)
Standard/Advanced 5000
4-bar Large Vessel Carrier Platform 24” (61cm)
24.4 x 24.3 x 14.2 (61.9 x 61.7 x 36.0cm)
Advanced 10000-1 / 10000-2
5-bar Large Vessel Carrier Platform
24” (61cm)
36.9 x 24.3 x 14.2 (93.7 x 61.7 x 36.0cm)
Advanced 15000-1 / 15000-2
VWR
®
Large Vessel Carrier Platforms
• Ideal for large sample containers like Carboys, jugs, and bottles
• High side walls secure samples
• Heavy-duty design
• Stainless steel construction
• Base tray has a non-skid rubber surface
Separatory Funnel Platform
• Holds 3 funnels at once on the included 18 x 18” (45.7 x 45.7cm) platform
• Accommodates 500mL to 2L separatory funnels
• Stainless steel construction
Description
Used on Shakers
Cat. No.
Separatory Funnel Platform
Standard/Advanced 3500, Advanced 3750
Replacement Part
Used with Carrier
Cat. No.
18” (45.7cm) adjustable bar (with mounting hardware)
24” (61cm) adjustable bar (with mounting hardware)
Microplate Clamp
• Holds one standard microplate or deep-well plate
• Constructed of type 304 stainless steel
VWR
®
SHAKERS
VWR
®
Platforms & Accessories
Description
Used on Shakers
Cat. No.
Microplate Clamp
Standard/Advanced 3500, 5000, 1000, 15000
Platform Capacities for Microplate Clamps
Platform
Size
Platform
Cat. No.
Microplate
Clamps
11 x 13
4
13 x 13
6
18 x 18
12
18 x 18
12
18 x 24
18
18 x 30
21
24 x 24
24
24 x 36
36
1...,28,29,30,31,32,33,34,35,36,37 39,40,41,42,43,44,45,46,47,48,...116
Powered by FlippingBook