VWR Collection Laboratory Equipment - page 37
35
Dedicated Platforms
• Pre-mounted flask clamps for maximum utilization of platform space for flasks of all one size
• Two-tier braces allow stacking of platforms with a 10” clearance (available for select sizes*)
• Platform is constructed of type 304 stainless steel
• Flask clamps are constructed of PVC
Platform Size / Flask Clamps Size
Flask Capacity
Used on Shakers
Cat. No.
13 x 13” (33 x 33cm) Dedicated Platform / 125mL Flask Clamp
16
Standard/Advanced 3500, Advanced 3750
13 x 13” (33 x 33cm) Dedicated Platform / 250mL Flask Clamp
12
Standard/Advanced 3500, Advanced 3750
13 x 13” (33 x 33cm) Dedicated Platform / 500mL Flask Clamp
8
Standard/Advanced 3500, Advanced 3750
13 x 13” (33 x 33cm) Dedicated Platform / 1L Flask Clamp
4
Standard/Advanced 3500, Advanced 3750
18 x 18” (45.7 x 45.7cm) Dedicated Platform / 125mL Flask Clamp *
27
Standard/Advanced 3500, Advanced 3750**
18 x 18” (45.7 x 45.7cm) Dedicated Platform / 250mL Flask Clamp *
20
Standard/Advanced 3500, Advanced 3750**
18 x 18” (45.7 x 45.7cm) Dedicated Platform / 500mL Flask Clamp *
13
Standard/Advanced 3500, Advanced 3750**
18 x 18” (45.7 x 45.7cm) Dedicated Platform / 1L Flask Clamp
9
Standard/Advanced 3500, Advanced 3750**
Two-Tier Braces (set of 4)
Standard/Advanced 3500
Adjustable Platforms
• Adjustable clamping bars accommodate various vessel types
• Stainless steel construction
• Base tray has a non-skid rubber surface
Description
Bar Size
Overall Dimensions
Used on Shakers
Cat. No.
2-bar Adjustable Platform
8.6” (21.8cm)
8.6 x 11.7” (21.8 x 29.7cm)
Mini Shaker, Mini Shaker 15
4-bar Adjustable Platform
18” (45.7cm)
18 x 18” (45.7 x 45.7cm)
Standard/Advanced 3500
4-bar Adjustable Platform
18” (45.7cm)
18 x 24” (45.7 x 61cm)
Standard/Advanced 5000
Replacement Part
Used with Platform
Cat. No.
18” (45.7cm) adjustable bar (with mounting hardware)
VWR
®
SHAKERS
VWR
®
Platforms
*Two tier ready
**Stacking of platforms is not recommended for Model 3750
1...,27,28,29,30,31,32,33,34,35,36 38,39,40,41,42,43,44,45,46,47,...116
Powered by FlippingBook