VWR Collection Laboratory Equipment - page 36
34
Universal Platforms
• Allows for mounting of flask clamps, test tube racks, and bottle clamps
• Platform slides over top of included tray and is tightened with adjustment screws
• Optional accessories screw directly into mounting point openings
• Two-tier braces allow stacking of platforms with a 10” clearance (available for select sizes*)
• Optional non-skid rubber mat can be placed on platform for an added non-slip surface
• Constructed of type 304 stainless steel
Description
Used on Shakers
Cat. No.
11 x 13” (27.9 x 33cm) Universal Platform
Standard/Advanced/Incubating 3500, Advanced 3750
13 x 13” (33 x 33cm) Universal Platform
Standard/Advanced 3500, Advanced 3750
18 x 18” (45.7 x 45.7cm) Universal Platform *
Standard/Advanced 3500, Advanced 3750 **
18 x 18” (45.7 x 45.7cm) Universal Platform
Incubating 5000I/5000IR
18 x 24” (45.7 x 61cm) Universal Platform *
Standard/Advanced 3500, Advanced 3750**, Standard/Advanced 5000**
24 x 24” (61 x 61cm) Universal Platform
Advanced 10000-1 / 10000-2
18 x 30” (45.7 x 76.2cm) Universal Platform *
Standard/Advanced 5000
24 x 36” (61 x 91.4cm) Universal Platform
Advanced 15000-1 / 15000-2
Two-Tier Braces (set of 4)
Standard/Advanced 3500, Standard/Advanced 5000
Description
Used on Shakers
Cat. No.
11 x 13” (27.9 x 33cm) Rubber Mat
Standard/Advanced/Incubating 3500, Advanced 3750
13 x 13” (33 x 33cm) Rubber Mat
Standard/Advanced 3500, Advanced 3750
18 x 18” (45.7 x 45.7cm) Rubber Mat
Standard/Advanced 3500, Advanced 3750
18 x 24” (45.7 x 61cm) Rubber Mat
Standard/Advanced 3500, Advanced 3750, Standard/Advanced 5000
24 x 24” (61 x 61cm) Rubber Mat
Advanced 10000-1 / 10000-2
24 x 36” (61 x 91.4cm) Rubber Mat
Advanced 15000-1 / 15000-2
Rubber Mats
Culture Platforms
• Ideal for slow speed applications; Petri dishes, culture flasks, and other flat bottom, low profile vessels
• Two-tier braces (available for both sizes) allow stacking of platforms with a 10” clearance
• Constructed of type 304 stainless steel
• Platform has a non-skid rubber surface
Description
Used on Shakers
Cat. No.
18 x 18” (45.7 x 45.7cm) Culture Platform *
Standard/Advanced 3500*, Advanced 3750**
18 x 24” (45.7 x 61cm) Culture Platform *
Standard/Advanced 3500*, Advanced 3750**, Standard/Advanced 5000
Two-Tier Braces (set of 4)
Standard/Advanced 3500, Standard/Advanced 5000
*Two tier ready
**Stacking of platforms is not recommended for Model 3750
VWR
®
SHAKERS
VWR
®
Platforms
1...,26,27,28,29,30,31,32,33,34,35 37,38,39,40,41,42,43,44,45,46,...116
Powered by FlippingBook