VWR Collection Laboratory Equipment - page 35
33
* Maximum speed/tilt angle may vary with heavy or unbalanced loads.
** Centered on tray.
Description
Electrical (50/60 Hz)
Cat. No.
Incubating 3-D Rotator Waver
120V | 5 amps | 450 watts
VWR
®
symphony™ Incubating 3-D Rotator Waber
• Electronic tilt adjustment from 0 to 20
°
while unit is operating
• LED displays for temperature, speed and tilt angle, and time
• Timer with audible alarm
• 2 year warranty
The VWR
®
symphony™ Incubating 3-D Rotator Waver combines the unique
vertical and horizontal “wave” motion with general purpose incubation in one
compact bench top unit.
Operating Features:
Microprocessor control:
The microprocessor control provides electronic tilt angle
adjustment which allows user to easily adjust waving angle from 0 to 20° while unit
is operating. Precise speed control provides smooth, low-speed waving motion
down to 1rpm.
PID temperature controller:
Maintains precise temperature control from ambient
+5°C to 65°C. Easy-to-use controls allow users to adjust temperature in 1°C
increments..
LED display:
Touch pad controls with easy-to-read, independent LED displays
for temperature, speed and tilt angle, and time allow operator to view all settings
at once. Provides repeatable and accurate results every time and is easily visible
across lab benches. Timer will display elapsed time or, when programmed to user
defined limit, will shut off unit when time reaches zero. Display will show last used
settings, even after power has been turned off.
Safety Features:
Overload protection:
Audible and visual signals will activate when system detects
an obstruction or overload of the tray.
Speed ramping feature:
Slowly increases speed to desired set-point to avoid
splashing.
Audible alarm:
In timed mode, alarm will sound when the time reaches zero.
Caution hot indicator:
Symbol illuminates when the temperature of the air in the
chamber reaches 40°C and remains lit until temperature cools down.
Spill-resistant design:
Channels fluids away from internal components.
Polycarbonate lid:
Permits viewing of samples without disturbing internal
temperature.
Operating Conditions:
Unit can be run in conditions from 5 to 40°C (41 to 104°F), maximum 80% relative
humidity, non-condensing.
Applications:
Cell cultures, hybridization procedures, and blotting techniques.
with
optional
26985-034
Ordering Information
Unit includes a 92” (234cm) detachable, 3-wire cord and plug. 2 year limited warranty on parts and labor.
Wave Motion
VWR
®
SHAKERS
VWR
®
symphony™ Incubating 3-D Rotator Waver
Specifications
Temperature Range
Ambient +5°C to 65°C
Temperature Uniformity
± 0.5°C at 37°C
Speed Range
1 to 30rpm*
Speed Accuracy
± 1rpm
Tilt Angle
0 to 20° *
Timer
1 second to 160 hours
Maximum Weight Capacity
5lbs (2.3kg)**
Tray Material
Aluminum
Tray Dimensions (L x W)
9.25 x 7.25” (23.5 x 18.4cm)
Interior Dimensions
10.75 x 7.75 x 3.4”
(27.3 x 19.7 x 8.6cm)
Overall Dimensions (L x W x H)
17 x 11 x 10.5”
(43.2 x 27.9 x 26.7cm)
Ship Weight
22lbs (10kg)
1...,25,26,27,28,29,30,31,32,33,34 36,37,38,39,40,41,42,43,44,45,...116
Powered by FlippingBook