VWR Collection Laboratory Equipment - page 33
31
Standard Test Tube Blocks
Sample Type
No. of Wells
Well Dia.
Well Depth
Cat. No.
6mm tube
30
8.3mm
48.4mm
10mm tube
24
10.7mm
48.4mm
12/13mm tube
20
13.9mm
48.4mm
Centrifuge Tube Combination Block
Sample Type
No. of Wells
Well Dia.
Well Depth
Cat. No.
1.5mL
4
11.1mm
39.1mm
15mL
3
17.1mm
44.5mm
50mL
2
29mm
47.6mm
Vial Blocks
Sample Type
No. of Wells
Well Dia.
Well Depth
Cat. No.
12mm vial
20
12.7mm
30mm
15mm vial
20
15.8mm
35mm
16mm vial
15
16.4mm
45mm
17mm vial
12
17.8mm
45mm
19mm vial
12
19.7mm
45mm
21mm vial
9
21.7mm
45mm
23mm vial
8
23.8mm
45mm
25mm vial
8
25.8mm
45mm
28mm vial
6
28.8mm
45mm
VWR
®
SHAKERS
VWR
®
symphony™ Incubating/Cooling Orbital Shaker Modular Blocks
1...,23,24,25,26,27,28,29,30,31,32 34,35,36,37,38,39,40,41,42,43,...116
Powered by FlippingBook