VWR Collection Laboratory Equipment - page 32
30
Tubes
Plates
Vials
Modular Blocks
VWR
®
Modular Blocks are constructed from a solid anodized aluminum block. The close contact of tubes-to-block-walls allow for maximum temperature
transfer.
Block dimensions (L x W x H):
3.75 x 3 x 2” (9.5 x 7.6 x 5.1cm)
Microcentrifuge Tube Blocks
Sample Type
No. of Wells Well Dia.
Well Depth
Cat. No.
0.5mL tube
30
7.9mm
27.6mm
1.5mL tube
20
11.1mm 39.1mm
2mL tube
20
11.5mm 38.1mm
Conical-Bottom Centrifuge Tube Blocks
Sample Type
No. of Wells
Well Dia.
Well Depth
Cat. No.
15mL tube
12
17.1mm 44.5mm
50mL tube
5
29mm
47.6mm
Applications:
Cell cultures, hybridizations, and extraction procedures.
VWR
®
SHAKERS
VWR
®
symphony™ Incubating/Cooling Orbital Shaker Modular Blocks
VWR modular blocks also fit in the VWR
®
Dry Block Heaters.
Constructed of anodized aluminum, this material is ideal for its temperature conducting and corrosion resistant properties.
1...,22,23,24,25,26,27,28,29,30,31 33,34,35,36,37,38,39,40,41,42,...116
Powered by FlippingBook