VWR Collection Laboratory Equipment - page 31
29
• Heats to 65°C and cools to 10°C below ambient
• LED displays for temperature, speed and time
• 2 year warranty
The VWR
®
symphony™ Incubating/Cooling Orbital Shaker is microplate ready
without the need for any additional accessories. Optional modular blocks can
accommodate micro-tubes, centrifuge tubes, vials, or culture tubes. Unit holds
microplates or modular blocks with a 5” (12.7cm) tall interior capacity. Ideal for
analysis that requires a stable, controlled temperature.
Operating Features:
Microprocessor control:
The variable speed microprocessor control provides
consistent uniform shaking action. Microprocessor will display last set-point and
will restart if power is interrupted.
PID temperature controller:
Maintains precise temperature control from 10°C
below ambient to 65°C. Easy-to-use controls allow users to adjust temperature in
1°C increments.
Triple eccentric drive:
Permanently lubricated ball bearings and maintenance
free, brushless DC motor provide reliable service and continuous duty operation.
LED display:
Touch pad controls with easy-to-read, independent LED displays
for temperature, speed, and time allow operator to view all settings at once.
Provides repeatable and accurate results every time and is easily visible across
lab benches. Timer will display elapsed time or, when programmed to user defined
limit, will shut off unit when time reaches zero. Display will show last used settings,
even after power has been turned off.
Temperature calibration mode:
Allows user to calibrate unit to an external
temperature device.
Safety Features:
Overload protection:
Audible and visual signals will activate when system detects
an obstruction or overload of the tray.
Speed ramping feature:
Slowly increases speed to desired set-point to avoid
splashing.
Audible alarm:
In timed mode, alarm will sound when the time reaches zero.
Alarm has optional mute function which can be set from the touch pad control.
Caution hot indicator:
Symbol illuminates when the temperature of the air in the
chamber reaches 40°C and remains lit until temperature cools down.
Spill-resistant design:
Channels fluids away from internal components.
Polycarbonate lid:
Permits viewing of samples without disturbing internal
temperature.
with optional
modular blocks
and
Operating Conditions:
Unit can be run in conditions from 5 to 40°C (41 to 104°F), maximum
80% relative humidity, non-condensing.
Applications:
Cell and bacterial cultures, hybridizations, and enzyme reactions.
Description
Electrical (50/60 Hz)
Cat. No.
Incubating/Cooling Orbital Shaker
120V | 2 amps | 200 watts
Incubating/Cooling Orbital Shaker
230V | 1 amps | 200 watts
VWR
®
symphony™ Incubating/Cooling Orbital Shaker
VWR
®
symphony™ Incubating/Cooling Orbital Shaker
Specifications
Temperature Range
10°C below ambient to 65°C
Temperature Uniformity
± 0.5°C at 37°C
Speed Range using Microplates
100 to 1200rpm
Speed Range using Modular Blocks
100 to 600rpm
Speed Accuracy
± 2%
Timer
1 second to 160 hours
Orbit
3mm (0.12”)
Maximum Weight Capacity
2 microplates or
2 modular blocks
Overall Dimensions (L x W x H)
17.9 x 11 x 10.5”
(45.5 x 27.9 x 26.7cm)
Ship Weight
34lbs (15.4kg)
Ordering Information
Unit includes a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). Units are also supplied with an adapter bracket
to hold optional modular blocks. See page 32 for block options. 2 year limited warranty on parts and labor.
VWR
®
SHAKERS
1...,21,22,23,24,25,26,27,28,29,30 32,33,34,35,36,37,38,39,40,41,...116
Powered by FlippingBook