VWR Collection Laboratory Equipment - page 29
27
VWR
®
SHAKERS
• LED displays for temperature, speed, and time
• Timer with audible alarm
• 2 year warranty
• Available with opaque lid for light sensitive samples
The VWR
®
symphony™
Incubating Microplate Shakers are optimized for shaking
microplates, deep-well plates, or micro-tubes.
Operating Features:
Microprocessor control:
The variable speedmicroprocessor control provides consistent
uniform shaking action. Microprocessor will display last set-point and will restart if power
is interrupted.
PID temperature controller:
Maintains precise temperature control from ambient
+5°C to 65°C. Easy-to-use controls allow users to adjust temperature in 1°C increments.
Triple eccentric drive:
Permanently lubricated ball bearings and maintenance free,
brushless DC motor provide reliable service and continuous duty operation.
LED display:
Touch pad controls with easy-to-read, independent LED displays for
temperature, speed, and time allow operator to view all settings at once. Provides
repeatable and accurate results every time and is easily visible across lab benches. Timer
will display elapsed time or, when programmed to user defined limit, will shut off unit when
time reaches zero. Display will show last used settings, even after power has been turned
off.
Temperature calibration mode:
Allows user to calibrate unit to an external temperature
device.
Safety Features:
Overload protection:
Audible and visual signals will activate when system detects an
obstruction or overload of the tray.
Speed ramping feature:
Slowly increases speed to desired set-point to avoid splashing.
Audible alarm:
In timed mode, alarm will sound when the time reaches zero.
Caution hot indicator:
Symbol illuminates when the temperature of the air in the
chamber reaches 40°C and remains lit until temperature cools down.
Spill-resistant design:
Channels fluids away from internal components.
Polycarbonate lid:
Clear lid permits viewing of samples without disturbing internal
temperature. Opaque lid prohibits light exposure to light sensitive samples.
Operating Conditions:
Unit can be run in conditions from 5 to 40°C (41 to 104°F), maximum 80% relative
humidity, non-condensing.
Applications:
Immunoassays and hybridizations.
Description
Electrical (50/60 Hz)
Cat. No.
Incubating Microplate Shaker
120V | 5 amps | 450 watts
Incubating Microplate Shaker
230V | 5 amps | 450 watts
Incubating Microplate Shaker with Opaque Lid
120V | 5 amps | 450 watts
Incubating Microplate Shaker with Opaque Lid
230V | 5 amps | 450 watts
VWR
®
symphony™ Incubating Microplate Shaker
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). 2 year limited warranty on parts and labor.
VWR
®
symphony™ Incubating Microplate Shaker
Specifications
Temperature Range
Ambient +5°C to 65°C
Temperature Uniformity
± 0.5°C at 37°C
Speed Range
100 to 1200rpm
Speed Accuracy
± 2%
Timer
1 second to 160 hours
Orbit
3mm (0.12”)
Maximum Weight Capacity
4 microplates or
2 micro-tube racks
Tray Material
Aluminum
Tray Dimensions (L x W)
11 x 7.75” (27.9 x 19.7cm)
Overall Dimensions (L x W x H)
17 x 11 x 7.75”
(43.2 x 27.9 x 19.7cm)
Ship Weight
30lbs (13.6kg)
with optional
1...,19,20,21,22,23,24,25,26,27,28 30,31,32,33,34,35,36,37,38,39,...116
Powered by FlippingBook