VWR Collection Laboratory Equipment - page 28
26
• LED displays for temperature, speed and time
• Timer with audible alarm
• 2 year warranty
The VWR
®
symphony™ Incubating Mini Shakers are designed to heat and shake
a variety of samples.
Operating Features:
Microprocessor control:
The variable speed microprocessor control provides
consistent uniform shaking action. Microprocessor will display last set-point and
will restart if power is interrupted.
PID temperature controller:
Maintains precise temperature control from ambient
+5°C to 65°C. Easy-to-use controls allow users to adjust temperature in 1°C
increments.
Triple eccentric drive:
Permanently lubricated ball bearings and maintenance
free, brushless DC motor provide reliable service and continuous duty operation.
LED display:
Touch pad controls with easy-to-read, independent LED displays for
temperature, speed and time allow operator to view all settings at once. Provides
repeatable and accurate results every time and is easily visible across lab benches.
Timer will display elapsed time or, when programmed to user defined limit, will shut
off unit when time reaches zero. Display will show last used settings, even after
power has been turned off.
Temperature calibration mode:
Allows user to calibrate unit to an external
temperature device.
Safety Features:
Overload protection:
Audible and visual signals will activate when system detects
an obstruction or overload of the tray.
Speed ramping feature:
Slowly increases speed to desired set-point to avoid
splashing.
Audible alarm:
In timed mode, alarm will sound when the time reaches zero.
Caution hot indicator:
Symbol illuminates when the temperature of the air in the
chamber reaches 40°C and remains lit until temperature cools down.
Spill-resistant design:
Channels fluids away from internal components.
Polycarbonate lid:
Permits viewing of samples without disturbing internal
temperature.
Operating Conditions:
Unit can be run in conditions from 5 to 40°C (41 to 104°F), maximum 80% relative
humidity, non-condensing.
Applications:
Cell cultures, bacterial suspensions, and hybridizations.
Ordering Information
Unit includes a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). 2 year limited warranty on parts and labor.
Description
Electrical (50/60 Hz)
Cat. No.
Incubating Mini Shaker
120V | 5 amps | 450 watts
Incubating Mini Shaker
230V | 5 amps | 450 watts
VWR
®
symphony™ Incubating Mini Shaker
VWR
®
SHAKERS
VWR
®
symphony™ Incubating Mini Shaker
Specifications
Temperature Range
Ambient +5°C to 65°C
Temperature Uniformity
± 0.5°C at 37°C
Speed Range
100 to 1200rpm
Speed Accuracy
± 2% of set speed
Timer
1 second to 160 hours
Orbit
3mm (0.12”)
Maximum Weight Capacity
8lbs (3.6kg)
Tray Material
Aluminum
Tray Dimensions (L x W)
11 x 7.75” (27.9 x 19.7cm)
Interior Dimensions
11.3 x 8.3 x 5.7”
(28.7 x 21.1 x 14.5cm)
Overall Dimensions (L x W x H)
17 x 11 x 10.7”
(43.2 x 27.9 x 27cm)
Ship Weight
30lbs (13.6kg)
1...,18,19,20,21,22,23,24,25,26,27 29,30,31,32,33,34,35,36,37,38,...116
Powered by FlippingBook