VWR Collection Laboratory Equipment - page 24
22
VWR
®
SHAKERS
• Shakes traditional or deep-well microplates
• Shakes up to 48 microplates at one time
The VWR
®
Advanced High Speed Microplate Shakers handle shaking for
difficult applications. These units operate at up to 2500rpm while holding deep-
well blocks and small diameter test tubes no taller than 5” (12.7cm).
Advanced High Speed Microplate Shaker has variable speed control from 600
to 2500rpm.
LED display:
Touch pad controls with easy-to-read, independent LED displays
for speed and time allow operator to view both settings at once. Provides
repeatable and accurate results every time and is easily visible across lab
benches. Timer will display elapsed time or when programmed to user defined
limit, will shut off unit when time reaches zero. Display will show last used
settings, even after power has been turned off.
Programmable “pulsing” feature:
Enhances shaking action and allows user
to program the unit to stop and start in a wide range of intervals.
RS-232 interface:
Provides two-way communication for data logging and unit
control
Operating Conditions:
Unit can be run in cold rooms and incubators from 4 to 40°C (39 to 104°F),
maximum 80% relative humidity, non-condensing.
Applications:
Emulsifications and cell lysing.
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). 2 year limited warranty on parts and labor.
Description
Electrical (50/60 Hz)
Cat. No.
Advanced High Speed Microplate Shaker
120V | 1 amp | 120 watts
Advanced High Speed Microplate Shaker
230V | 1 amp | 120 watts
VWR
®
Advanced High Speed Microplate Shaker
VWR
®
Advanced High Speed Microplate Shaker
Specifications
Speed Range
600 to 2500rpm
Timer
1 second to 160 hours
Orbit
3.6mm (0.14”)
Maximum Weight Capacity
48 microplates or up to 7lbs (3.2kg)
Tray Material
Aluminum
Tray Dimensions (L x W)
12.25 x 11” (31.1 x 27.9cm)
Overall Dimensions (L x W x H)
15.5 x 12 x 12.8”
(39.4 x 30.5 x 32.5cm)
Ship Weight
50lbs (22.7kg)
1...,14,15,16,17,18,19,20,21,22,23 25,26,27,28,29,30,31,32,33,34,...116
Powered by FlippingBook