VWR Collection Laboratory Equipment - page 22
20
VWR
®
SHAKERS
• Electronic tilt adjustment from 0 to 15° while unit is operating
• Displays for speed and tilt angle, and time
• Timer with audible alarm
• 2 year warranty
The VWR
®
Rocking Platform Shaker is ideal for cell culture work and is designed to
be used in a variety of environmental conditions.
Operating Features:
Low profile design:
Takes up less space and easily fits into most fume hoods and
incubators. Cast aluminum base offers durability and added stability.
Microprocessor control:
The microprocessor control provides electronic tilt angle
adjustment which allows user to easily adjust rocking angle from 0 to 15° while unit
is operating. Precise speed control provides smooth, low-speed rocking motion
down to 1rpm.
LED display:
Touch pad controls with easy-to-read, independent LED displays for
speed and tilt angle, and time allow operator to view all settings at once. Provides
repeatable and accurate results every time and is easily visible across lab benches.
Timer will display elapsed time or, when programmed to user defined limit, will shut
off unit when time reaches zero. Display will show last used settings, even after
power has been turned off.
Safety Features:
Overload protection:
Audible and visual signals will activate when system detects
an obstruction or overload of the tray.
Speed ramping feature:
Slowly increases speed to desired set-point to avoid
splashing.
Audible alarm:
In timed mode, alarm will sound when the time reaches zero.
Spill-resistant design:
Channels fluids away from internal components.
Operating Conditions:
Unit can be run in cold rooms, incubators, and CO
2
environments from -10 to 60°C
(14 to 140°F), maximum 80% relative humidity, non-condensing.
Applications:
Staining and destaining gels, hybridization procedures, hematology, and blotting
techniques.
* Maximum speed/tilt angle may vary with heavy or unbalanced loads.
** Centered on tray.
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). Units are also supplied with a 12.75 x 10”
(32.4 x 25.4cm) non-skid rubber mat. 2 year limited warranty on parts and labor.
Description
Electrical (50/60 Hz)
Cat. No.
Rocking Platform Shaker
120V | 5 amps | 25 watts
Rocking Platform Shaker
230V | 5 amps | 25 watts
VWR
®
Rocking Platform Shaker
Rocking Motion
VWR
®
Rocking Platform Shaker
Specifications
Speed Range
1 to 50rpm*
Speed Accuracy
± 1rpm
Tilt Angle
0 to 15° *
Timer
1 second to 160 hours
Maximum Weight Capacity
10lbs (4.5kg)**
Tray Material
Aluminum
Tray Dimensions (L x W)
12.75 x 10” (32.4 x 25.4cm)
Overall Dimensions (L x W x H)
17 x 11 x 5”
(43.2 x 27.9 x 12.7cm)
Ship Weight
15.5lbs (7kg)
1...,12,13,14,15,16,17,18,19,20,21 23,24,25,26,27,28,29,30,31,32,...116
Powered by FlippingBook