VWR Collection Laboratory Equipment - page 21
19
VWR
®
SHAKERS
• Holds up to 4 microplates or 2 micro-tube racks
• Accepts deep well plates
• Timer with audible alarm
The VWR
®
Microplate Shaker is ideal for immunoassays and general microplate
shaking, and is designed to be used in a variety of environmental conditions.
Operating Features:
Low profile design:
Takes up less space and easily fits into most hoods and
incubators. Cast aluminum base offers durability and added stability.
Microprocessor control:
The variable speed microprocessor control provides
consistent uniform shaking action.
Triple eccentric drive:
Permanently lubricated ball bearings and maintenance
free, brushless DC motor provide reliable service and continuous duty operation.
LED display:
Touch pad controls with easy-to-read, independent LED displays for
speed and time allow operator to view all settings at once. Provides repeatable
and accurate results every time and is easily visible across lab benches. Timer will
display elapsed time or when programmed to user defined limit, will shut off unit
when time reaches zero. Display will show last used settings, even after power has
been turned off.
Safety Features:
Overload protection:
Audible and visual signals will activate when system detects
an obstruction or overload of the tray.
Speed ramping feature:
Slowly increases speed to desired set-point to avoid
splashing.
Audible alarm:
In timed mode, alarm will sound when the time reaches zero.
Spill-resistant design:
Channels fluids away from internal components.
Operating Conditions:
Unit can be run in cold rooms, incubators, and CO
2
environments from -10 to 60°C
(14 to 140°F), maximum 80% relative humidity, non-condensing.
Applications:
ELISA assays and DNA studies.
Description
Electrical (50/60 Hz)
Cat. No.
Microplate Shaker
120V | 5 amps | 25 watts
Microplate Shaker
230V | 5 amps | 25 watts
VWR
®
Microplate Shaker
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). 2 year limited warranty on parts and labor.
VWR
®
Microplate Shaker
Specifications
Speed Range
100 to 1200rpm
Speed Accuracy
± 2%
Timer
1 second to 160 hours
Orbit
3mm (0.12”)
Maximum Weight Capacity
4 microplates or 2 micro-tube racks
Tray Material
Aluminum
Tray Dimensions (L x W)
11 x 7.75” (27.9 x 19.7cm)
Overall Dimensions (L x W x
H)
17 x 11 x 4”
(43.2 x 27.9 x 10.2cm)
Ship Weight
25lbs (11.3kg)
1...,11,12,13,14,15,16,17,18,19,20 22,23,24,25,26,27,28,29,30,31,...116
Powered by FlippingBook