VWR Collection Laboratory Equipment - page 20
18
VWR
®
SHAKERS
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). Units are also supplied with an 11.75 x 8.75”
(29.9 x 22cm) non-skid rubber mat. 2 year limited warranty on parts and labor.
Description
Electrical (50/60 Hz)
Cat. No.
Mini Shaker
120V | 5 amps | 25 watts
Mini Shaker
230V | 5 amps | 25 watts
Mini Shaker 15
120V | 5 amps | 25 watts
Mini Shaker 15
230V | 5 amps | 25 watts
VWR
®
Mini Shaker and Mini Shaker 15
with
optional flask
clamps
• General purpose shaker
• LED displays for speed and time
• 3mm or 15mm orbit
• 2 year warranty
VWR
®
Mini Shaker and Mini Shaker 15 are ideal for a wide variety of shaking
applications. Tray includes a non-skid rubber mat. Remove the mat to mount a
variety of optional flask clamps or test tube racks directly onto the tray.
Operating Features:
Low profile design:
Takes up less space and easily fits into most hoods and
incubators. Cast aluminum base offers durability and added stability.
Microprocessor control:
The variable speed microprocessor control provides
consistent uniform shaking action.
Triple eccentric drive:
Permanently lubricated ball bearings and maintenance
free, brushless DC motor provide reliable service and continuous duty operation.
LED display:
Touch pad controls with easy-to-read, independent LED displays for
speed and time allow operator to view both settings at once. Provides repeatable
and accurate results every time and is easily visible across lab benches. Timer will
display elapsed time or, when programmed to user defined limit, will shut off unit
when time reaches zero. Display will show last used settings, even after power
has been turned off.
Safety Features:
Overload protection:
Audible and visual signals will activate when system detects
an obstruction or overload of the tray.
Speed ramping feature:
Slowly increases speed to desired set-point to avoid
splashing.
Audible alarm:
In timed mode, alarm will sound when the time reaches zero.
Spill-resistant design:
Channels fluids away from internal components.
Operating Conditions:
Units can be run in cold rooms, incubators, and CO
2
environments.
Model 1000-3: -10 to 60°C (14 to 140°F)
Model 1000-15: -10 to 40°C (14 to 104°F)
Maximum 80% relative humidity, non-condensing.
Applications:
Cell cultures, immunoassays, and protein studies.
VWR
®
Mini Shaker and Mini Shaker 15
Specifications
Speed Range
1000-3
100 to 1200rpm
1000-15
40 to 300rpm
Speed Accuracy
± 2%
Timer
1 second to 160 hours
Orbit
1000-3
3mm (0.12”)
1000-15
15mm (0.6”)
Maximum Weight Capacity
8lbs (3.6kg)
Tray Material
Aluminum
Tray Dimensions (L x W)
11.75 x 8.75” (29.9 x 22cm)
Overall Dimensions (L x W x H)
17 x 11 x 4”
(43.2 x 27.9 x 10.2cm)
Ship Weight
25lbs (11.3kg)
1...,10,11,12,13,14,15,16,17,18,19 21,22,23,24,25,26,27,28,29,30,...116
Powered by FlippingBook