VWR Collection Laboratory Equipment - page 19
17
VWR
®
SHAKERS
• Variable speed microprocessor control
• Low profile design
The VWR
®
Standard 1000 Orbital Shaker is an economical shaker designed for
educational labs or basic shaking applications.
Operating Features:
Low profile design:
Takes up less space and easily fits into most hoods and
incubators. Cast aluminum base offers durability and added stability.
Microprocessor control:
The variable speed microprocessor control provides
consistent uniform shaking action.
Triple eccentric drive:
Permanently lubricated ball bearings and maintenance
free, brushless DC motor provide reliable service and continuous duty operation.
Adjustment knob:
Basic speed knob with dial settings from 1 to 10.
Safety Features:
Speed ramping feature:
Slowly increases speed to desired set-point to avoid
splashing.
Spill-resistant design:
Channels fluids away from internal components.
Operating Conditions:
Unit can be run in cold rooms, incubators, and CO
2
environments from 0 to 40°C (32
to 104°F), maximum 80% relative humidity, non-condensing.
Applications:
Blotting techniques, staining/destaining, and general shaking procedures.
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). Units are also supplied with an 11.75 x 8.75” (29.9
x 22.2cm) non-skid rubber mat. 2 year limited warranty on parts and labor.
VWR
®
Standard 1000 Orbital Shaker
VWR
®
Standard 1000 Orbital Shaker
Specifications
Speed Range
40 to 300rpm
Orbit
15mm (0.6”)
Maximum Weight Capacity
8lbs (3.6kg)
Tray Material
Aluminum
Tray Dimensions (L x W)
11.75 x 8.75” (29.9 x 22.2cm)
Overall Dimensions (L x W x H)
17 x 11 x 4”
(43.2 x 27.9 x 10.2cm)
Ship Weight
25lbs (11.3kg)
Description
Electrical (50/60 Hz)
Cat. No.
Standard 1000 Orbital Shaker
120V | 5 amps | 25 watts
Standard 1000 Orbital Shaker
230V | 2.5 amps | 25 watts
1...,9,10,11,12,13,14,15,16,17,18 20,21,22,23,24,25,26,27,28,29,...116
Powered by FlippingBook