VWR Collection Laboratory Equipment - page 18
16
VWR
®
SHAKERS
• Exclusive Accu-drive Shaking System*
• Available with either 1” or 2” orbit
• 5 year warranty on motor and drive mechanism
VWR
®
Advanced 15000-1 and 15000-2 Orbital Shakers are the largest capacity
shakers designed for the heaviest of loads. They have the most powerful drive
mechanism of all the shakers and large orbits for optimal shaking of larger vessels.
Operating Features:
Accu-Drive Shaking System*:
The exclusive Accu-Drive Shaking System delivers
exceptional speed control, accuracy, and durability. The system continuously
monitors shaking speed and maintains set-point, even under changing loads.
When unit is set to a speed above 100rpm, the speed accuracy is ± 1% of set
speed. When set under 100rpm, speed accuracy is ± 1rpm.
Microprocessor control:
The variable speed microprocessor control provides
consistent uniform shaking action.
Triple eccentric drive:
Permanently lubricated ball bearings and maintenance
free, brushless DC motor provide reliable service and continuous duty operation.
LED display:
Touch pad controls with easy-to-read, independent LED displays for
speed and time allow operator to view both settings at once. Provides repeatable
and accurate results every time and is easily visible across lab benches. Timer will
display elapsed time or, when programmed to user defined limit, will shut off unit
when time reaches zero. Display will show last used settings, even after power has
been turned off.
RS-232 interface:
Provides two-way communication for data logging and unit
control.
Speed calibration mode:
Allows user to automatically recalibrate speed display.
Safety Features:
Load sensor:
A built-in load sensor detects unbalanced conditions and
automatically reduces rpm to a safe speed to protect samples.
Overload protection:
Audible and visual signals will activate when system detects
an obstruction or overload of the tray.
Speed ramping feature:
Slowly increases speed to desired set-point to avoid
splashing.
Audible alarm:
In timed mode, alarm will sound when the time reaches zero.
Alarm has optional mute function which can be set from the touch pad control.
Spill-resistant design:
Channels fluids away from internal components.
Operating Conditions:
Units can be run in cold rooms, incubators, and CO
2
environments from -10 to 60°C
(14 to 140°F), maximum 80% relative humidity, non-condensing.
Applications:
Cell cultures, solubility studies, and extraction procedures.
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt
units are supplied with Euro type plug). Units are also supplied with a 24
x 36” (61 x 91cm) non-skid rubber mat. 2 year limited warranty on parts
and labor, 5 year limited warranty on motor and drive mechanis
m.
* Patent Pending
Description
Electrical (50/60 Hz)
Cat. No.
Advanced 15000-1 Orbital Shaker
120V | 5 amps | 80 watts
Advanced 15000-1 Orbital Shaker
230V | 2.5 amps | 80 watts
Advanced 15000-2 Orbital Shaker
120V | 5 amps | 80 watts
Advanced 15000-2 Orbital Shaker
230V | 2.5 amps | 80 watts
VWR
®
Advanced 15000 Orbital Shaker
with optional
platform
and
flask clamps
with
optional platform
VWR
®
Advanced 15000 Orbital Shakers
Specifications
Speed Range
15000-1
15 to 500rpm
15000-2
15 to 300rpm
Speed Accuracy
above 100rpm ± 1% of set speed
below 100rpm ± 1rpm
Timer
1 second to 160 hours
Orbit
15000-1
25mm (1”)
15000-2
51mm (2”)
Maximum Weight Capacity
150lbs (68kg)
Tray Material
Aluminum
Tray Dimensions (L x W)
24 x 36” (61 x 91cm)
Overall Dimensions (L x W x H)
28.25 x 26.7 x 7”
(71.8 x 67.8 x 17.8cm)
Ship Weight
230lbs (104.4kg)
1...,8,9,10,11,12,13,14,15,16,17 19,20,21,22,23,24,25,26,27,28,...116
Powered by FlippingBook