VWR Collection Laboratory Equipment - page 16
14
VWR
®
SHAKERS
• LED displays for speed and time
• Exclusive Accu-Drive Shaking System*
• 50lb weight capacity
• 5 year warranty on motor and drive mechanism
The VWR
®
Advanced 5000 Orbital Shaker is designed for a wide range of
applications with larger or heavier loads that require accurate and repeatable
results.
Operating Features:
Accu-Drive Shaking System*:
The exclusive Accu-Drive Shaking System
delivers exceptional speed control, accuracy, and durability. The shaking system
continuously monitors shaking speed and maintains set-point, even under
changing loads. When unit is set to a speed above 100rpm, the speed accuracy is
± 1% of set speed. When set under 100rpm, speed accuracy is ± 1rpm.
Microprocessor control:
The variable speed microprocessor control provides
consistent uniform shaking action.
Triple eccentric drive:
Permanently lubricated ball bearings and maintenance
free, brushless DC motor provide reliable service and continuous duty operation.
LED display:
Touch pad controls with easy-to-read, independent LED displays for
speed and time allow operator to view both settings at once. Provides repeatable
and accurate results every time and is easily visible across lab benches. Timer
will display elapsed time or, when programmed to user defined limit, will shut
off unit when time reaches zero. Display will show last used settings, even after
power has been turned off.
RS-232 interface:
Provides two-way communication for data logging and unit
control.
Speed calibration mode:
Allows user to automatically recalibrate speed display.
Safety Features:
Load sensor:
A built-in load sensor detects unbalanced conditions and
automatically reduces rpm to a safe speed to protect samples.
Overload protection:
Audible and visual signals will activate when system
detects an obstruction or overload of the tray.
Speed ramping feature:
Slowly increases speed to desired set-point to avoid
splashing.
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). Units are also supplied with an 18 x 24” (45.7 x 61cm)
non-skid rubber mat. 2 year limited warranty on parts and labor, 5 year limited warranty on motor and drive mechanism.
Description
Electrical (50/60 Hz)
Cat. No.
Advanced 5000 Orbital Shaker
120V | 5 amps | 75 watts
Advanced 5000 Orbital Shaker
230V | 2.5 amps | 75 watts
VWR
®
Advanced 5000 Orbital Shaker
with optional
platform
and
flask clamps
Audible alarm:
In timed mode, alarm will sound when the time
reaches zero. Alarm has optional mute function which can be set from
the touch pad control.
Spill-resistant design:
Channels fluids away from internal
components.
Operating Conditions:
Unit can be run in cold rooms, incubators, and CO
2
environments
from -10 to 60°C (14 to 140°F), maximum 80% relative humidity, non-
condensing.
Applications:
Cell cultures, solubility studies, and extraction procedures.
* Patent Pending
VWR
®
Advanced 5000 Orbital Shaker
Specifications
Speed Range
20 to 500rpm
Speed Accuracy
above 100rpm ± 1% of set speed
below 100rpm ± 1rpm
Timer
1 second to 160 hours
Orbit
25mm (1”)
Maximum Weight Capacity
50lbs (22.7kg)
Tray Material
Aluminum
Tray Dimensions (L x W)
18 x 24” (45.7 x 61cm)
Overall Dimensions (L x W x H)
23.6 x 26.7 x 6”
(59.9 x 67.8 x 15.2cm)
Ship Weight
109lbs (49.5kg)
1...,6,7,8,9,10,11,12,13,14,15 17,18,19,20,21,22,23,24,25,26,...116
Powered by FlippingBook