VWR Collection Laboratory Equipment - page 15
13
VWR
®
SHAKERS
• Microprocessor controls
• Continuous or timed operation
• 50lb weight capacity
The VWR
®
Standard 5000 Orbital Shaker is designed for applications with
heavy duty loads. Shaker provides reproducible motion that is evenly distributed
throughout the entire surface of the tray.
Operating Features:
Microprocessor control:
The variable speed microprocessor control provides
consistent uniform shaking action.
Triple eccentric drive:
Permanently lubricated ball bearings and maintenance
free, brushless DC motor provide reliable service and continuous duty operation.
Adjustment knobs:
Basic speed and time knobs with dial settings from 1 to 10.
Safety Features:
Speed ramping feature:
Slowly increases speed to desired set-point to avoid
splashing.
Spill-resistant design:
Channels fluids away from internal components.
Operating Conditions:
Unit can be run in cold rooms, incubators, and CO
2
environments from 0 to 40°C
(32 to 104°F), maximum 80% relative humidity, non-condensing.
Applications:
Bacterial suspensions, staining/destaining, and general mixing.
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). Units are also supplied with an 18 x 24” (45.7 x 61cm)
non-skid rubber mat. 2 year limited warranty on parts and labor.
Description
Electrical (50/60 Hz)
Cat. No.
Standard 5000 Orbital Shaker
120V | 5 amps | 75 watts
Standard 5000 Orbital Shaker
230V | 2.5 amps | 75 watts
VWR
®
Standard 5000 Orbital Shaker
with
optional platform
VWR
®
Standard 5000 Orbital Shaker
Specifications
Speed Range
25 to 500rpm
Timer
1 minute to 120 minutes
Orbit
25mm (1”)
Maximum Weight Capacity
50lbs (22.7kg)
Tray Material
Aluminum
Tray Dimensions (L x W)
18 x 24” (45.7 x 61cm)
Overall Dimensions (L x W x H)
23.5 x 26.7 x 6”
(59.9 x 67.8 x 15.2cm)
Ship Weight
109lbs (49.5kg)
1...,5,6,7,8,9,10,11,12,13,14 16,17,18,19,20,21,22,23,24,25,...116
Powered by FlippingBook