VWR Collection Laboratory Equipment - page 12
10
VWR
®
SHAKERS
• Microprocessor controls
• Continuous or timed operation
• 35lb weight capacity
The VWR
®
Standard 3500 Orbital Shaker is designed for a wide range of
applications that require basic shaking control. Shaker provides reproducible
motion that is evenly distributed throughout the entire surface of the tray.
Operating Features:
Microprocessor control:
The variable speed microprocessor control provides
consistent uniform shaking action.
Triple eccentric drive:
Permanently lubricated ball bearings and maintenance
free, brushless DC motor provide reliable service and continuous duty operation.
Adjustment knobs:
Basic speed and time knobs with dial settings from 1 to 10.
Safety Features:
Speed ramping feature:
Slowly increases speed to desired set-point to avoid
splashing.
Spill-resistant design:
Channels fluids away from internal components.
Operating Conditions:
Unit can be run in cold rooms, incubators, and CO
2
environments from 0 to 40°C
(32 to 104°F), maximum 80% relative humidity, non-condensing.
Applications:
Bacterial suspensions, staining/destaining, and general mixing.
Ordering Information
Unit includes a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). Unit is also supplied with an 11 x 13” (27.9 x
33cm) non-skid rubber mat. 2 year limited warranty on parts and labor.
Description
Electrical (50/60 Hz)
Cat. No.
Standard 3500 Orbital Shaker
120V | 5 amps | 75 watts
Standard 3500 Orbital Shaker
230V | 2.5 amps | 75 watts
VWR
®
Standard 3500 Orbital Shaker
with optional
platform
and
flask clamps
&
Specifications
Speed Range
25 to 500rpm
Timer
1 minute to 120 minutes
Orbit
19mm (0.75”)
Maximum Weight Capacity
35lbs (16kg)
Tray Material
Aluminum
Tray Dimensions (L x W)
11 x 13” (27.9 x 33cm)
Overall Dimensions (L x W x H)
16.25 x 14 x 5.75”
(41.3 x 35.6 x 14.6cm)
Ship Weight
49lbs (22.2kg)
VWR
®
Standard 3500 Orbital Shaker
1...,2,3,4,5,6,7,8,9,10,11 13,14,15,16,17,18,19,20,21,22,...116
Powered by FlippingBook