VWR Collection Laboratory Equipment - page 115
113
VWR
®
Opti-Flow
VWR
®
Opti-Flow Forced Ventilation System*
Opti-Flow Forced Ventilation System*
Fan(s) draw air from the chamber and baffles deflect air around the sides of the chamber. This prevents the flow of air directly on samples delivering
exceptional temperature uniformity and stability.
This feature can be found on the VWR® symphony™ Incubating 3500 Orbital Shaker, Incubating 5000I Orbital Shaker, and Incubating/Refrigerating
5000IR Orbital Shaker (see pages 23-25).
* Patent Pending
1...,105,106,107,108,109,110,111,112,113,114 116
Powered by FlippingBook