VWR Collection Laboratory Equipment - page 113
111
OTHER QUALITY VWR
®
PRODUCTS
Other Quality VWR
®
Products
VWR
®
Inoculating Turntables
VWR
®
hand-operated inoculating turntables produce almost concentric circles of
bacterial colonies that are evenly distributed across petri dishes. Cast iron turntables
are coated with gray baked acrylic enamel.
The 76mm (3”) high, small turntable accommodates 100mm petri dishes. It has a
tripod base to bring work approximately to eye level. It has a center disk covered
with non-skid rubber lining.
The large turntable accommodates 100mm or 150mm petri dishes. It has a height of
only 32mm (1.26”), ideal for steadying a forearm on the work surface while plating.
The large turntable contains two circular sections. The top section has a raised
gripper lip as well as a recessed center disk covered with non-skid rubber lining.
VWR
®
Slide Staining Rack
Constructed of stainless steel, the VWR
®
Slide Staining Rack will resist corrosion
under normal use. Adjustable to fit trays or sinks up to 533mm (21”) inside.
Description
Dimensions (L x W)
Cat. No.
Slide Staining Rack
603 x 89mm (23.74 x 3.5”)
VWR
®
Airejector
Rapid air removal evacuates 1 liter of air to 711mm (28”) of vacuum in 30 seconds.
Works with compressed air sources to 60 psi. Nickel-plated construction.
Description
Length
Cat. No.
Airejector
140mm (5.51”)
Description
Dimensions (Dia. x H)
Cat. No.
Small Turntable
114 x 77mm (4.49 x 3.03”)
Large Turntable
150 x 32mm (5.91 x 1.26”)
1...,103,104,105,106,107,108,109,110,111,112 114,115,116
Powered by FlippingBook