VWR Collection Laboratory Equipment - page 112
110
VWR
®
Microplate Stability Chamber
VWR
®
MICROPLATE STABILITY CHAMBER
The Microplate Stability Chamber is a tool proven to reduce the variation often
experienced in cell-based assays. This chamber is inserted into standard or CO
2
incubators. Traditional incubators often create heterogeneous environments
for cell growth and biochemical assays due to frequent door openings and
“hot spots” within the incubator. This chamber uses the incubator heat and
atmosphere to create a homogeneous micro-environment for microwell plates.
High humidity within the chamber eliminates evaporation from the plates that
causes the edge effect. The thick aluminum shelf distributes heat evenly to the
plates thus creating excellent temperature uniformity. Variation in cell-based
assays has been reduced by as much as 50% using the Microplate Stability
Chamber. The chamber includes a fan driven by the appropriate power adapter.
Overall dimensions (L x W x H): 16 x 14 x 3.1” (40.6 x 35.6 x 7.9cm)
Tray Dimensions (L x W H):
13.75 x 11.2 x 0.75” (34.9 x 28.4 x 1.9cm)
Microplate Stability Tray
The shelf used in the Microplate Stability Chamber can improve temperature
uniformity of microwell plates in incubators. The anodized aluminum shelf holds
six plates. One included with unit.
Description
Cat. No.
Microplate Stability Chamber, 120V (50/60 Hz)
Microplate Tray
1...,102,103,104,105,106,107,108,109,110,111 113,114,115,116
Powered by FlippingBook