VWR Collection Laboratory Equipment - page 11
9
Incubating
Mini Shaker
Incubating
Microplate Shaker
Incubating/Cooling Shaker
for PCR
Incubating/Cooling
Orbital Shaker
Incubating Rocking
Platform Shaker
Incubating 3-D
Rotator Waver
Ambient +5° to 65°C
Ambient +5° to 65°C 10°C Below Ambient to 65°C 10°C Below Ambient to 65°C Ambient +5° to 65°C
Ambient +5° to 65°C
100 to 1200rpm
100 to 1200rpm
100 to 1600rpm
100 to 1200rpm
1 to 50rpm
1 to 30rpm
1 second to 160 hours
1 second to 160 hours
up to 160 hours
1 second to 160 hours
1 second to 160 hours
1 second to 160 hours
Orbital
Orbital
Orbital
Orbital
Rocking
Waving
3mm
3mm
3mm
3mm
Tilt Angle: 0 to 15° *
Tilt Angle: 0 to 20° *
8lbs
4 Microplates /
2 Micro-Tube Racks
2 PCR Plates
2 Microplates / 2 Modular
Blocks
10lbs**
5lbs**
YES
YES
YES
YES
YES
YES
NO
NO
NO
NO
NO
NO
Triple Eccentric
Triple Eccentric
Triple Eccentric
Triple Eccentric
Cable
Cable
Brushless DC Motor
Brushless DC Motor
Brushless DC Motor
Brushless DC Motor
Stepper Motor
Stepper Motor
YES - Displays Last Setting YES - Displays Last Setting YES - Displays Last Setting YES - Displays Last Setting YES - Displays Last Setting YES - Displays Last Setting
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
NO
NO
NO
NO
NO
NO
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
NO
NO
NO
NO
NO
NO
NO
NO
NO
NO
NO
NO
NO
NO
2 year
2 year
2 year
2 year
2 year
2 year
17 x 11 x 10.5”
17 x 11 x 7.75”
17.9 x 11 x 10.5”
17.9 x 11 x 10.5”
17 x 11 x 10.5”
17 x 11 x 10.5”
11 x 7.75”
11 x 7.75”
N/A
N/A
10 x 7.5”
9.25 x 7.25”
N/A
N/A
N/A
N/A
N/A
N/A
VWR
®
Incubating Shaker Selection Guide
* Maximum speed/tilt angle may vary with heavy or unbalanced loads
** Centered on tray.
VWR
®
SHAKERS
1...,2,3,4,5,6,7,8,9,10 12,13,14,15,16,17,18,19,20,21,...116
Powered by FlippingBook