VWR Collection Laboratory Equipment - page 109
107
VWR
®
Heavy-Duty Lab-Jacks
• Stainless steel construction
• Seven convenient sizes to choose from
• Autoclavable and chemical resistant
These heavy-duty, stainless steel Lab-Jacks are ultra-stable lifting platforms with
exceptional strength and durability. Constructed of 300 series stainless steel, Lab-Jacks
are designed for use in extreme environments and high load applications. Equipped
with over-sized, sure-grip adjustment knobs that provide extra leverage for easy height
adjustments. Durable construction allows lifts to be autoclaved or chemically cleaned.
Ideal for use in fume hoods or bench tops and hold a variety of items such as glassware,
hotplates, baths, and magnetic stirrers.
Deck Size
Min. to Max. Height
Max. Load*
Cat. No.
3 x 3” (76 x 76mm)
2.5 to 5” (64 to 127mm)
20lbs (9.08kg)
4 x 4” (102 x 102mm)
2.5 to 5” (64 to 127mm)
30lbs (13.62kg)
6 x 6” (152 x 152mm)
3 to 9.75” (76 to 248mm)
35lbs (15.89kg)
8 x 8” (203 x 203mm)
3.5 to 13” (89 to 330mm)
55lbs (24.97kg)
10 x 10” (254 x 254mm)
3.5 to 13” (89 to 330mm)
80lbs (36.32kg)
12 x 12” (305 x 305mm)
4 to 19.5” (102 to 495mm)
150lbs (68.1kg)
16 x 16” (406 x 406mm)
4 to 19.5” (102 to 495mm) 180lbs (81.72kg)
Description
Rod Diameter
Cat. No.
17” Support Rod Kit
0.51” (13mm)
Ratchet Tool
N/A
* Maximum load rating represents static weight only. Static weight is the amount a unit can hold, not lift.
Optional Accessories
VWR
®
17” Support Rod Kit
Ideal for creating an adjustable support stand for mounting various items such as thermometer
clamps, temperature probes, flask and column clamps. This kit allows you to mount a 17”
(432mm) threaded vertical support rod to the upper deck of a 6 x 6”, 8 x 8”, 10 x 10”, 12 x
12”, or 16 x 16” Lab-Jacks by screwing the rod into the pre-drilled hole.
17” Support Rod Kit includes:
1 - 17” (432mm) stainless steel rod
1 - jam nut
1 - flat washer
Ratchet Tool
Designed to add extra leverage to your 12 x 12” or 16 x 16” Lab-Jack. This recommended
Ratchet Tool easily attaches to the actuating rod to allow easy, accurate adjustments.
VWR
®
Lab-Lifts
1...,99,100,101,102,103,104,105,106,107,108 110,111,112,113,114,115,116
Powered by FlippingBook