VWR Collection Laboratory Equipment - page 107
105
VWR
®
CLAMPS
Description
Dimensions L x W x H
Cylinder Diameter
Cat. No.
Model 701 PVC Coated Stand
17.25 x 17.25 x 10.75” (438 x 438 x 273mm)
7 to 10” (178 x 254mm)
VWR
®
Model 701 PVC Coated Stand
The PVC coated stand has a flat bottom support so it can’t “ride up” on the
cylinder. Includes an adjustable 1” (25mm) wide, 54” (1372mm) long nylon strap
with nickel-plated, non-slip spring catch and buckle for fast, easy adjustment to
keep the cylinder safely in place against upper support bar. Lower support bar
stops cylinder shift. The PVC coating prevents marring of the cylinder surface.
This clamp is not available with a safety message strap.
VWR
®
Cylinder Stands
Description
Dimensions (Dia. x H)
Cylinder Diameter
Cat. No.
Model 704 Adjustable Stand
18.5 x 7.5” (470 x 191mm)
6 to 9.25” (152 x 235mm)
VWR
®
Model 704 Adjustable Stand
Heavy-duty, cast aluminum stand with adjustable “L” brackets hold cylinders from
6 to 9.25” (152 to 235mm) in diameter. Hinges open for installation without lifting
the cylinder or disturbing connections. Prevents accidental tipping. Hinges open
for easy, no-lifting installation.
Description
Dimensions (Dia. x H)
Cylinder Diameter
Cat. No.
Model 703 Stand
18.25 x 6” (464 x 152mm)
8 to 9” (203 x 229mm)
VWR
®
Model 703 Stand
This cast aluminum stand hinges open and has 4 thumbscrews to tightly grip
cylinders from 8 to 9” (203 to 229mm) in diameter.
Description
Dimensions (Dia. x H)
Cylinder Diameter
Cat. No.
Model 713 Portable Stand
15.5 x 3” (394 x 76mm)
7 to 9.25” (178 x 235mm)
VWR
®
Model 713 Portable Stand
This cast iron portable cylinder stand offers a sturdy cylinder stand and a
convenient cylinder dolly all-in-one. This cylinder stand is designed in two
interlocking halves and can be installed without lifting the tank. Built-in rubber
wheels for easy mobility. Three thumbscrews tightly grip cylinder.
VWR
®
Gas Cylinder Safety Supports
1...,97,98,99,100,101,102,103,104,105,106 108,109,110,111,112,113,114,115,116
Powered by FlippingBook