VWR Collection Laboratory Equipment - page 106
104
VWR
®
CLAMPS
Description
Dimensions L x W x H
Cylinder Diameter
Cat. No.
Model 715 Wall Bracket with Strap
1.875 x 8.125 x 4.625” (48 x 206 x 118mm)
4 to 14” (102 to 356mm)
Model 715 Wall Bracket with Safety Message Strap
1.875 x 8.125 x 4.625” (48 x 206 x 118mm)
4 to 14” (102 to 356mm)
VWR
®
Model 715 Wall Bracket
Cylinder wall-mount brackets are constructed of cast aluminum and contoured to allow
cylinders to fit firmly along the support edge. Recessed screw holes at each side of the
bracket allow for easy wall mounting. Features a 1” (25mm) wide, 54” (1372mm) long
nylon strap with nickel-plated, non-slip spring catch and buckle for fast, easy adjustment.
Available with or without a “Secure Cylinder” safety message strap. For cylinders from 4
to 14” (102 to 356mm) in diameter.
Description
Dimensions L x W x H
Cylinder Diameter
Cat. No.
Model 717 Wall Bracket with Strap & Chain
1.875 x 8.125 x 4.625” (48 x 206 x 118mm)
4 to 14” (102 to 356mm)
Model 717 Wall Bracket with Safety Message Strap & Chain 1.875 x 8.125 x 4.625” (48 x 206 x 118mm)
4 to 14” (102 to 356mm)
safety message strap
safety message strap
VWR
®
Model 717 Wall Bracket
This wall bracket is similar to Model 715, but includes a sturdy safety chain for extra
security. Chain measures 49“ (124.5cm)
VWR
®
Cylinder Wall Brackets
VWR
®
Gas Cylinder Safety Supports
1...,96,97,98,99,100,101,102,103,104,105 107,108,109,110,111,112,113,114,115,...116
Powered by FlippingBook