VWR Collection Laboratory Equipment - page 105
103
VWR
®
CLAMPS
VWR
®
Cylinder Bench Clamps
Rugged cast aluminum clamps safely secure gas cylinders to benches, tables or other flat surfaces up to 2.5” (64mm) thick. The 1” (25mm) wide, 54” (1372mm)
long nylon strap features a nickel-plated, non-slip spring catch and buckle for easy adjustment around cylinders from 4 to 14” (102 to 356mm) in diameter. Models
711 and 716 available with or without a “Secure Cylinder” safety message strap.
Description
Dimensions L x W x H (closed)
Cylinder Diameter
Cat. No.
Model 711 Bench Clamp with Strap
3.25 x 5.25 x 6.5” (83 x 133 x 165mm)
4 to 14” (102 to 356mm)
Model 711 Bench Clamp with Safety Message Strap
3.25 x 5.25 x 6.5” (83 x 133 x 165mm)
4 to 14” (102 to 356mm)
Description
Dimensions L x W x H (closed)
Cylinder Diameter
Cat. No.
Model 716 Bench Clamp with Strap & Chain
3.25 x 5.25 x 6.5” (83 x 133 x 165mm)
4 to 14” (102 to 356mm)
Model 716 Bench Clamp with Safety Message Strap & Chain 3.25 x 5.25 x 6.5” (83 x 133 x 165mm)
4 to 14” (102 to 356mm)
VWR
®
Model 716 Bench Clamp
This bench clamp is similar to Model 711, but includes a sturdy safety chain for extra
security. Chain measures 49 “ (124.5cm)
Description
Dimensions L x W x H (closed)
Cylinder Diameter
Cat. No.
Model 712 Heavy-Duty Bench Clamp with Strap
3.25 x 6 x 4.5” (83 x 152 x 114mm)
4 to 14” (102 to 356mm)
VWR
®
Model 712 Heavy-Duty Bench Clamp
This bench clamp has two screw clamps to tighten for extra firm mounting to any flat
surface up to 1.75” (45mm) thick with a 1.25” (32mm) overhang. Especially convenient
for temporary storage situations. Rugged cast aluminum bench clamp features a 1” (25mm)
wide, 54” (1372mm) long nylon strap with nickel-plated, non-slip spring catch buckle to hold
cylinders from 4 to 14” (102 to 356mm) in diameter. This clamp is not available with a
safety message strap.
safety message strap
safety message strap
VWR
®
Model 711 Bench Clamp
This bench clamp has a large tightening handle for mounting to any flat surface up to 2.5”
(64mm) thick. A nylon pad prevents damage to the bench or table surface. Two tapered
mounting screw holes are provided for permanent attachment to bench-top.
VWR
®
Gas Cylinder Safety Supports
1...,95,96,97,98,99,100,101,102,103,104 106,107,108,109,110,111,112,113,114,115,...116
Powered by FlippingBook