VWR Collection Laboratory Equipment - page 104
102
VWR
®
Support Plates
Designed to hold Hotplates/Stirrers and other apparatus to lab-frames or ring stands.
Aluminum construction offers strength and durability. Support plates include a non-skid
rubber mat and a built-in holder that grips rods up to 0.75” (19mm) in diameter.
Plate Diameter
Distance from Rod
Cat. No.
6” (152mm)
2.6” (66mm)
VWR
®
Round Support Plate
Aluminum plate supports beakers, mantles and petri dishes. Fits standard lab-frames using
VWR
®
connectors or holders, shown on pages 93-95.
Size
Plate Dimensions
Distance from Rod
Cat. No.
Small
9.5 x 6.5” (241 x 165mm)
1.9” (48mm)
Medium
12.75 x 9” (324 x 229mm)
1.9” (48mm)
Large
15.75 x 12” (400 x 305mm)
1.9” (48mm)
VWR
®
Talon
®
Support Plates
VWR
®
CLAMPS
1...,94,95,96,97,98,99,100,101,102,103 105,106,107,108,109,110,111,112,113,114,...116
Powered by FlippingBook