VWR Collection Laboratory Equipment - page 103
101
VWR
®
CLAMPS
Cast iron support stand bases are constructed with black enamel finish for
chemical resistance and durability. All bases feature a built-in support rod holder
with locking knob that accepts a 0.51” (13mm) diameter support rod. U-shaped
base features three built-in support rod holders.
* Support rods are not included.
VWR
®
Tripod Base Support Stand, cast iron
VWR
®
U-Shaped Base Support Stand, cast iron
Description
Diameter
Cat. No.
12” (305mm) Aluminum Rod
0.51” (13mm)
18” (457mm) Aluminum Rod
0.51” (13mm)
24” (610mm) Aluminum Rod
0.51” (13mm)
36” (914mm) Aluminum Rod
0.51” (13mm)
VWR
®
Rectangular Base Support Stand, cast iron
Description
Dimensions
Cat. No.
Rectangular Base (only)
4 x 6” (102 x 152mm)
Rectangular Base (only)
5 x 8” (127 x 203mm)
Rectangular Base (only)
6 x 9” (152 x 229mm)
Rectangular Base (only)
8 x 10” (203 x 254mm)
Description
Overall Footprint Diameter
Cat. No.
Tripod Base (only)
14.75” (375mm)
Description
Dimensions
Cat. No.
U-Shaped Base (only)
7 x 7” (178 x 178mm)
VWR
®
Aluminum Support Rods
Description
Diameter
Cat. No.
12” (305mm) Stainless Steel Rod
0.51” (13mm)
18” (457mm) Stainless Steel Rod
0.51” (13mm)
24” (610mm) Stainless Steel Rod
0.51” (13mm)
36” (914mm) Stainless Steel Rod
0.51” (13mm)
VWR
®
Stainless Steel Support Rods
VWR
®
Talon
®
Support Stands
1...,93,94,95,96,97,98,99,100,101,102 104,105,106,107,108,109,110,111,112,113,...116
Powered by FlippingBook