VWR Collection Laboratory Equipment - page 102
100
VWR
®
Support Stands
This Support Stand is made entirely of 303 stainless steel. It consists of a
sturdy base that can accommodate vessels up to 18” (457mm) in diameter within
the “U” shaped base. The threaded stainless steel support rod is 0.63” (16mm)
in diameter and is screwed into the support stand base. The base measures
17.75” (451mm) across and 10.5” (267mm) deep overall.
* Includes support rod.
Description
Cat. No.
Support Stand with 23” (584mm) Rod
Support Stand with 28” (711mm) Rod
Support Stand with 36” (914mm) Rod
Support Stand with 40” (1016mm) Rod
Support Stand with 48” (1219mm) Rod
Support Stand with 60” (1524mm) Rod
VWR
®
Heavy-Duty Support Stands
Heavy-Duty Support Stand was designed to permit cylindrical vessels up to 12”
(305mm) in diameter to sit close to the support rod. Support rod is ideal for
mounting stirring motors for heavy equipment. Each of the four corners rests on
a rubber cushion. The cast iron base measures 16.5” (419mm) across and 12”
(305mm) deep overall and has a black enamel finish.
* Includes support rod.
Description
Cat. No.
Heavy-Duty Support Stand with 18” (457mm) Rod
Heavy-Duty Support Stand with 23” (584mm) Rod
Heavy-Duty Support Stand with 28” (711mm) Rod
Heavy-Duty Support Stand with 36” (914mm) Rod
Heavy-Duty Support Stand with 40” (1016mm) Rod
Heavy-Duty Support Stand with 48” (12119mm) Rod
Heavy-Duty Support Stand with 60” (1524mm) Rod
Support Stand
Heavy-Duty Support Stand
Description
Cat. No.
Support Stand Stabilizer Knobs (2 per pack)
VWR
®
Support Stand Stabilizer Knobs
Support Stand Stabilizer Knobs can be added to the two additional threaded
holes in the base legs to secure the stand to a bench top.
VWR
®
Replacement Support Rods
Description
Diameter
Cat. No.
18” (45.7cm) stainless steel rod
0.63” (16mm)
23” (58.4cm) stainless steel rod
0.63” (16mm)
28” (71.1cm) stainless steel rod
0.63” (16mm)
36” (91.4cm) stainless steel rod
0.63” (16mm)
40” (101.6cm) stainless steel rod
0.63” (16mm)
48” (121.9cm) stainless steel rod
0.63” (16mm)
60” (152.4cm) stainless steel rod
0.63” (16mm)
VWR
®
Talon
®
Support Stands
VWR
®
CLAMPS
1...,92,93,94,95,96,97,98,99,100,101 103,104,105,106,107,108,109,110,111,112,...116
Powered by FlippingBook