VWR Collection Laboratory Equipment - page 101
99
VWR
®
CLAMPS
VWR
®
Heavy-Duty Lab-Frame
Floor mounted, free-standing unit is specifically designed for heavy-duty work
such as pilot plant set-ups. Outside frame is constructed of a strong, but light
weight steel channel. Interior lattice uses a horizontal channel connector; one
end accommodates the rod, the other end fits into the channel and locks tightly
with a bolt. Loosen the bolt, and the connector slides easily along the channel
track for simple lattice adjustment. Entire apparatus is supported on sturdy,
counter-weighted cast iron feet, which may be bolted to the floor for added
stability required in top-heavy installations.
Components: (5) 48” (1219mm) rods
(3) 72” (1829mm) rods
(15) S-Connectors
(16) Channel Connectors
(24) clamping nuts
(2) 21.7” (551mm) cast iron feet
(1) frame kit
(1) wrench
VWR
®
Fume Hood Kits
Choose from four kits specifically designed to fit within laboratory fume hoods.
Rods are constructed of stainless steel.
4’ (1.22m) Fume Hood Kit
Components: (8) 2” (51mm) rods
(2) 12” (305mm) rods
(2) 18” (457mm) rods
(4) 36” (914mm) rods
(4) 38” (965mm) rods
(30) Frame Connectors
(4) Rod End Connectors
(4) Lab-Frame Feet
Frame: 36 x 38” (914 x 965mm)
Base: 12” (305mm) wide
5’ (1.52m) Fume Hood Kit
Components: (8) 2” (51mm) rods
(2) 12” (305mm) rods
(2) 18” (457mm) rods
(5) 36” (914mm) rods
(4) 50” (1270mm) rods
(34) Frame Connectors
(4) Rod End Connectors
(4) Lab-Frame Feet
Frame: 36 x 50” (914 x 1270mm)
Base: 12” (305mm) wide
6’ (1.83m) Fume Hood Kit
Components: (8) 2” (51mm) rods
(2) 12” (305mm) rods
(2) 18” (457mm) rods
(6) 36” (914mm) rods
(4) 62” (1575mm) rods
(38) Frame Connectors
(4) Rod End Connectors
(4) Lab-Frame Feet
Frame: 36 x 62” (914 x 1575mm)
Base: 12” (305mm) wide
8’ (2.44m) Fume Hood Kit
Components:(10) 2” (51mm) rods
(3) 12” (305mm) rods
(2) 18” (457mm) rods
(8) 36” (914mm) rods
(4) 86” (2184mm) rods
(45) Frame Connectors
(4) Rod End Connectors
(6) Lab-Frame Feet
Frame: 36 x 86” (914 x 2184mm)
Base: 12” (305mm) wide
VWR
®
Talon
®
Lab-Frames
Frame Dimensions
Overall Dimensions
Aluminum
Cat. No.
Fiberglass
Cat. No.
Stainless Steel
Cat. No.
48 x 72” (1219 x 1829mm)
75 x 72” (1905 x 1829mm)
Cat. No.
Cat. No.
Cat. No.
Cat. No.
14224-400
1...,91,92,93,94,95,96,97,98,99,100 102,103,104,105,106,107,108,109,110,111,...116
Powered by FlippingBook