VWR Collection Laboratory Equipment - page 100
98
VWR
®
Large Lab-Frame
This 48 x 48” (1219 x 1219mm) frame is ideal for complex glassware setups. Base
is 18” (457mm) wide for stability.
Components: (10) 2” (51mm) rods
(3) 18” (457mm) rods
(2) 36” (914mm) rods
(10) 48” (1219mm) rods
(38) S-Connectors
(4) Rod End Connectors
(6) Lab-Frame Feet
VWR
®
Extra-Large Lab-Frame
Versatile, adaptable Extra-Large Lab-Frame measures 48 x 72” (1219 x 1829mm)
and may be used either horizontally or vertically.
Components: (6) 2” (51mm) rods
(6) 12” (305mm) rods
(2) 24” (610mm) rods
(7) 48” (1219mm) rods
(5) 72” (1829mm) rods
(45) S-Connectors
(4) Rod End Connectors
(8) Lab-Frame Feet
VWR
®
Talon
®
Lab-Frames
VWR
®
CLAMPS
Frame Dimensions
Aluminum
Cat. No.
Fiberglass
Cat. No.
Stainless Steel
Cat. No.
48 x 72” (1219 x 1829mm)
Frame Dimensions
Base Dimensions
Aluminum
Cat. No.
Fiberglass
Cat. No.
Stainless Steel
Cat. No.
48 x 48” (1219 x 1219mm)
18” (457mm) wide
1...,90,91,92,93,94,95,96,97,98,99 101,102,103,104,105,106,107,108,109,110,...116
Powered by FlippingBook