VWR Collection Laboratory Equipment - page 10
8
Model
Thermal Shake
Touch
Cooling Thermal
Shake Touch
Incubating 3500
Orbital Shaker
Incubating 5000I
Orbital Shaker
Incubating/Refrigerating
5000IR Orbital Shaker
Temperature Range
4°C above ambient to 100°C 13°C below ambient to 100°C Ambient +5° to 65°C
Ambient +5° to 65°C 15°C Below Ambient to 65°C
Speed Range
300 to 3000rpm
300 to 3000rpm
15 to 500rpm
15 to 500rpm
15 to 500rpm
Timer
1 minute to 99 hours, 59 min.
1 minute to 99 hours, 59 min.
1 second to 160 hours
1 second to 160 hours
1 second to 160 hours
Motion
Orbital
Orbital
Orbital
Orbital
Orbital
Orbit
3mm
3mm
19mm
25.4mm
25.4mm
Maximum Weight Capacity
1 Block
1 Block
35lbs
50lbs
50lbs
Audible Alarm
YES
YES
YES
YES
YES
Load Sensor
NO
NO
YES
YES
YES
Drive System
N/A
N/A
Triple Eccentric
Triple Eccentric
Triple Eccentric
Motor Type
Brushless DC Motor
Brushless DC Motor
Brushless DC Motor
Brushless DC Motor
Brushless DC Motor
Set-Point Retention
YES - Displays Last Setting YES - Displays Last Setting YES - Displays Last Setting YES - Displays Last Setting YES - Displays Last Setting
Restart / Power Out
YES
YES
YES
YES
YES
Ramp to Speed
YES
YES
YES
YES
YES
Temperature Overshoot
Protection
NO
NO
YES
YES
YES
Overload Protection
NO
NO
YES
YES
YES
User Calibration
(Temperature)
YES
YES
YES
YES
YES
User Calibration (Speed)
NO
NO
YES
YES
YES
Interface
USB
USB
RS-232
RS-232
RS-232
Warranty
2 years
2 years
2 / 5 year
2 / 5 Year
2 / 5 Year
Overall Dimensions
(L x W x H)
10.25 x 9.75 x 5.2”
10.25 x 9.75 x 5.2”
25.4 x 14 x 16”
32.1 x 26.6 x 23.5”
41.1 x 26.6 x 23.5”
Included Tray (L x W)
NO
NO
13 x 11”
18 x 18”
18 x 18”
Platform Options (L x W)
N/A
N/A
13 x 11”
18 x 18”
18 x 18”
VWR
®
Incubating Shaker Selection Guide
VWR
®
SHAKERS
1,2,3,4,5,6,7,8,9 11,12,13,14,15,16,17,18,19,20,...116
Powered by FlippingBook