BDH Chemicals 2013 - page 84
Sulfuric Acid, BDH ARISTAR® PLUS
High purity acid for quantitative trace metal analysis at the parts-per-billion
(ppb) level.
Assay (H
2
SO
4
, w/w)........................................................................................................................ 93 - 98%
Color.....................................................................................................................................10 APHA max.
Aluminum (Al)............................................................................................................................ 1 ppb max.
Antimony (Sb)............................................................................................................................ 1 ppb max.
Arsenic (As)............................................................................................................................ 0.5 ppb max.
Barium (Ba)............................................................................................................................ 0.1 ppb max.
Beryllium (Be)......................................................................................................................... 0.1 ppb max.
Bismuth (Bi)............................................................................................................................ 0.1 ppb max.
Cadmium (Cd)......................................................................................................................... 0.5 ppb max.
Calcium (Ca).............................................................................................................................. 1 ppb max.
Cerium (Ce)............................................................................................................................. 0.1 ppb max.
Cesium (Cs)............................................................................................................................. 0.1 ppb max.
Chromium (Cr)......................................................................................................................... 0.5 ppb max.
Cobalt (Co).............................................................................................................................. 0.5 ppb max.
Copper (Cu)............................................................................................................................. 0.5 ppb max.
Dysprosium (Dy)...................................................................................................................... 0.1 ppb max.
Erbium (Er)............................................................................................................................. 0.1 ppb max.
Europium (Eu)......................................................................................................................... 0.1 ppb max.
Gadolinium (Gd)...................................................................................................................... 0.1 ppb max.
Gallium (Ga)........................................................................................................................... 0.1 ppb max.
Germanium (Ge)........................................................................................................................ 1 ppb max.
Gold (Au)................................................................................................................................ 0.5 ppb max.
Hafnium (Hf)........................................................................................................................... 0.1 ppb max.
Holmium (Ho)......................................................................................................................... 0.1 ppb max.
Indium (In).............................................................................................................................. 0.1 ppb max.
Iron (Fe).................................................................................................................................... 1 ppb max.
Lanthanum (La)....................................................................................................................... 0.1 ppb max.
Lead (Pb)................................................................................................................................ 0.1 ppb max.
Lithium (Li)............................................................................................................................. 0.5 ppb max.
Lutetium (Lu).......................................................................................................................... 0.1 ppb max.
Magnesium (Mg)........................................................................................................................ 1 ppb max.
Manganese (Mn)..................................................................................................................... 0.5 ppb max.
Mercury (Hg)........................................................................................................................... 0.1 ppb max.
Molybdenum (Mo)................................................................................................................... 0.5 ppb max.
Neodymium (Nd)..................................................................................................................... 0.1 ppb max.
Nickel (Ni).............................................................................................................................. 0.5 ppb max.
Niobium (Nb).......................................................................................................................... 0.1 ppb max.
Palladium (Pd)....................................................................................................................Information only
Platinum (Pt).......................................................................................................................Information only
Potassium (K)............................................................................................................................. 1 ppb max.
Praseodymium (Pr).................................................................................................................. 0.1 ppb max.
Rhodium (Rh).......................................................................................................................... 0.5 ppb max.
Rubidium (Rb)......................................................................................................................... 0.5 ppb max.
Samarium (Sm)....................................................................................................................... 0.1 ppb max.
Scandium (Sc)......................................................................................................................... 0.1 ppb max.
Selenium (Se)........................................................................................................................... 10 ppb max.
Silver (Ag).................................................................................................................................. 1 ppb max.
Sodium (Na)............................................................................................................................... 1 ppb max.
Strontium (Sr).......................................................................................................................... 0.5 ppb max.
Tantalum (Ta)......................................................................................................................Information only
Tellurium (Te).......................................................................................................................... 0.1 ppb max.
Terbium (Tb)........................................................................................................................... 0.1 ppb max.
Thallium (Tl)........................................................................................................................... 0.1 ppb max.
Thorium (Th)........................................................................................................................... 0.1 ppb max.
Thulium (Tm).......................................................................................................................... 0.1 ppb max.
Tin (Sn)..................................................................................................................................... 1 ppb max.
Titanium (Ti).............................................................................................................................. 1 ppb max.
Tungsten (W).......................................................................................................................... 0.5 ppb max.
Uranium (U)............................................................................................................................ 0.1 ppb max.
Vanadium (V).......................................................................................................................... 0.5 ppb max.
Ytterbium (Yb)........................................................................................................................ 0.1 ppb max.
Yttrium (Y).............................................................................................................................. 0.1 ppb max.
Zinc (Zn).................................................................................................................................... 1 ppb max.
Zirconium (Zr)......................................................................................................................... 0.5 ppb max.
Chloride (Cl-)...........................................................................................................................0.7 ppm max.
Total Phosphorus (P)..............................................................................................................0.05 ppm max.
Nitrate (NO
3
)........................................................................................................................... 0.2ppm max.
Substances reducing permanganate (KMnO
4
).............................................................................. 20 ppm max.
Size
Packaging
Cat. No.
Price
500 mL
Poly Bottle
Ea./
89.89
2.5 L
Poly Bottle
Ea./
158.27
SULFURIC ACID 50%
Formula:
H₂SO₄
MW:
98.08 g/mol
Boiling Pt:
Min. 100°C (1013 hPa)
Density:
1.1 to 1.4 g/cm³ (20°C)
CAS:
7664-93-9
Sulfuric Acid, Solution. 50%, v/v (1+1).
APHA.
Appearance/color................................................................................................................................Clear
Shelf-life..........................................................................................................................................2 years
Size
Packaging
Cat. No.
Price
500 mL
HDPE Bottle
Ea./
27.34
Cs. 12/
311.63
4 L
HDPE Bottle
Ea./
125.51
Cs. 4/
502.03
EINECS:
231-639-5
ADR:
8,II
Index:
016-020-00-8
UN:
2796
SULFURIC ACID 25% (V/V)
Formula:
H
2
SO
4
MW:
98.08 g/mol
CAS:
7664-93-9
EINECS:
231-639-5
Sulfuric Acid, Solution. 25% (v/v).
Made with deionized water.
Appearance/color................................................................................................................................Clear
Shelf-life..........................................................................................................................................2 years
Density............................................................................................................................ 1.24-1.27 at 25 °C
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
33.08
Cs. 12/
377.14
SULFURIC ACID 20%
Formula:
H₂SO₄
MW:
98.08 g/mol
CAS:
7664-93-9
EINECS:
231-639-5
Sulfuric Acid, Solution. 20%, v/v (1+4).
Made with deionized water
Appearance/color................................................................................................................................Clear
Shelf-life..........................................................................................................................................2 years
Density.....................................................................................................................1.2031-1.2353 at 20 °C
Sulfuric acid (v/v).......................................................................................................................... 19.9-20.1
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
25.63
Cs. 12/
292.14
4 L
Poly Bottle
Ea./
57.03
Cs. 4/
227.45
ADR:
8,II
Index:
016-020-00-8
UN:
2796
ADR:
8,II
Index:
016-020-00-8
UN:
2796
OH
OH
O O S
OH
OH
O O S
OH
OH
O O S
66
|
BDH Chemicals
|
us.vwr.com/bdh
|
800.932.5000
S
Sulfuric Acid
1...,74,75,76,77,78,79,80,81,82,83 85,86,87,88,89,90,91,92,93,94,...100
Powered by FlippingBook