BDH Chemicals 2013 - page 78
SODIUM HYDROXIDE 0.05 MOL/L
(0.05 N) AQUEOUS SOLUTION
Formula:
NaOH
MW:
40 g/mol
CAS:
1310-73-2
Sodium Hydroxide, Solution. 0.05N.
AOAC, APHA, and ASTM.
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Normality............................................................................................................................. 0.0495-0.0505
Shelf-life........................................................................................................................................... 1 year
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
21.68
Cs. 12/
247.17
SODIUM HYDROXIDE 0.02 MOL/L
(0.02 N) AQUEOUS SOLUTION
Formula:
NaOH
MW:
40 g/mol
Density:
1 g/cm³ (20°C)
CAS:
1310-73-2
EINECS:
215-185-5
ADR:
8,III
UN:
1824
Sodium Hydroxide, Solution. 0.02N.
AOAC, APHA, and ASTM for acidity.
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Normality............................................................................................................................. 0.0195-0.0205
Shelf-life........................................................................................................................................... 1 year
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
19.97
Cs. 12/
227.67
SODIUM HYDROXIDE 0.01 MOL/L
(0.01 N) AQUEOUS SOLUTION
Formula:
NaOH
MW:
40 g/mol
CAS:
1310-73-2
EINECS:
215-185-5
ADR:
8,III
UN:
1824
Sodium Hydroxide, Solution. 0.01N.
Made with deionized water. Suitable as a titrant.
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Normality............................................................................................................................. 0.0095-0.0105
Shelf-life........................................................................................................................................... 1 year
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
30.58
Cs. 12/
348.61
EINECS:
215-185-5
ADR:
8,III
UN:
1824
SODIUM HYDROXIDE,
0.25N AQUEOUS SOLUTION
Formula:
NaOH
MW:
40 g/mol
Density:
1.04 g/cm³ (25°C)
CAS:
1310-73-2
EINECS:
215-185-5
ADR:
8,II
UN:
1824
Sodium Hydroxide, Solution. 0.25N.
ASTM.
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Normality............................................................................................................................. 0.2495-0.2505
Shelf-life..........................................................................................................................................2 years
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
19.03
Cs. 12/
216.99
4 L
Poly Bottle
Ea./
42.11
Cs. 4/
169.08
SODIUM HYDROXIDE 0.2 MOL/L (0.2 N)
AQUEOUS SOLUTION
Formula:
NaOH
MW:
40 g/mol
Density:
1.01 g/cm³ (25°C)
CAS:
1310-73-2
Sodium Hydroxide, Solution. 0.2N.
AOAC, APHA, and ASTM.
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Normality............................................................................................................................. 0.1995-0.2005
Shelf-life.....................................................................................................................................18 months
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
22.11
Cs. 12/
252.09
4 L
Poly Bottle
Ea./
47.58
Cs. 4/
189.36
EINECS:
215-185-5
ADR:
8,III
UN:
1824
SODIUM HYDROXIDE, 0.1N
AQUEOUS SOLUTION
Formula:
NaOH
MW:
40 g/mol
Density:
1 g/cm³ (25°C)
CAS:
1310-73-2
Sodium Hydroxide, Solution. 0.1N.
AOAC, APHA, and ASTM.
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Normality............................................................................................................................. 0.0995-0.1005
Shelf-life..........................................................................................................................................2 years
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
31.57
Cs. 12/
359.89
4 L
Poly Bottle
Ea./
72.67
Cs. 4/
262.87
4 L
Cubitainer®
Ea./
51.13
Cs. 4/
194.29
20 L
Cubitainer®
Ea./
172.72
EINECS:
215-185-5
ADR:
8,III
UN:
1824
NaOH
NaOH
NaOH
NaOH
NaOH
NaOH
60
|
BDH Chemicals
|
us.vwr.com/bdh
|
800.932.5000
S
Sodium Hydroxide
1...,68,69,70,71,72,73,74,75,76,77 79,80,81,82,83,84,85,86,87,88,...100
Powered by FlippingBook