BDH Chemicals 2013 - page 76
SODIUM HYDROXIDE, 5N
AQUEOUS SOLUTION
Formula:
NaOH
MW:
40 g/mol
Boiling Pt:
109°C (1013 hPa)
Density:
1.327 g/cm³ (20°C)
Sodium Hydroxide, Solution. 5.000N.
Made with deionized water. Suitable as a titrant.
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Normality.................................................................................................................................... 4.99-5.01
Shelf-life..........................................................................................................................................2 years
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
26.04
Cs. 12/
296.84
4 L
Poly Bottle
Ea./
71.92
Cs. 4/
289.37
10 L
Cubitainer®
Ea./
100.04
SODIUM HYDROXIDE 3 MOL/L
(3 N) AQUEOUS SOLUTION
Formula:
NaOH
MW:
40 g/mol
CAS:
1310-73-2
Sodium Hydroxide, Solution. 3.0N.
Made with deionized water
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Normality.................................................................................................................................2.995-3.005
Shelf-life..........................................................................................................................................2 years
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
33.03
Cs. 12/
376.56
CAS:
1310-73-2
EINECS:
215-185-5
ADR:
8,II
UN:
1824
EINECS:
215-185-5
ADR:
8,II
UN:
1824
NaOH
NaOH
SODIUM HYDROXIDE, 6N
AQUEOUS SOLUTION
Formula:
NaOH
MW:
40 g/mol
Density:
1.22 g/cm³ (20°C)
CAS:
1310-73-2
Sodium Hydroxide, Solution. 6.0N.
APHA.
Appearance/color................................................................................................................................Clear
N.I.S.T. traceable.................................................................................................................................... Yes
Normality.................................................................................................................................... 5.97-6.03
Shelf-life..........................................................................................................................................2 years
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
34.87
Cs. 12/
397.47
EINECS:
215-185-5
ADR:
8,II
UN:
1824
NaOH
VWR® sympHony™ Benchtop and Portable Meters
A Complete Line of Electrochemistry Meters for
All your Testing and Measurement Needs
Meters offer a user-friendly design and interface with step-by-step on screen prompts. Provide consistently accurate and
reliable measurements. All meters can be configured to meet your specific needs. Includes VWR three-year limited warranty.
Description
Cat. No.
B10C Meter - Conductivity
89231-676
B20PI Meter - pH, ISE
89231-692
B30PCI Meter - pH, Conductivity, ISE
89231-696
B40PCID Meter - pH, Conductivity, ISE, Dissolved Oxygen
89231-684
H30PCD Portable Meter - pH, Conductivity, Dissolved Oxygen
89231-688
H30PCO Portable Meter - pH, Conductivity, ORP
89231-700
For additional meters and accessories, visit
us.vwr.com
58
|
BDH Chemicals
|
us.vwr.com/bdh
|
800.932.5000
S
Sodium Hydroxide
1...,66,67,68,69,70,71,72,73,74,75 77,78,79,80,81,82,83,84,85,86,...100
Powered by FlippingBook