BDH Chemicals 2013 - page 71
SILVER NITRATE, SOLUTION, 0.0192N
Silver Nitrate, Solution. 0.0192N.
Normality........................................................................................................................... 0.0187 - 0.0197
Appearance/color.................................................................................................................................Pass
Size
Packaging
Cat. No.
Price
1 L
Amber Glass Bottle
Ea./
40.88
Cs. 12/
466.03
SILVER NITRATE, SOLUTION, 0.0141N
Silver Nitrate, Solution. 0.0141N.
Size
Packaging
Cat. No.
Price
1 L
HDPE Bottle
Ea./
87.97
Cs. 12/
1002.88
SODIUM ACETATE TRIHYDRATE
Acetic acid sodium salt trihydrate
Formula:
H₃CCOONa•
3
H₂O
MW:
136.08 g/mol
Boiling Pt:
120 to 123 °C (1013 hPa)
Melting Pt:
58°C
Density:
1.4 g/cm³ (20°C)
CAS:
6131-90-4
EINECS:
204-823-8
Sodium Acetate Trihydrate, ACS
Colorless or white solid
Assay...................................................................................................................99 – 101% NaC
2
H
3
O
2
·3H
2
O
pH of a solution at 25.0°C................................................................................................................7.5 – 9.2
Substances reducing permanganate..............................................................................................Passes test
Insoluble matter....................................................................................................................... 0.005% max
Chloride (Cl)............................................................................................................................. 0.001% max
Phosphate (PO
4
)......................................................................................................................... 5 ppm max
Sulfate (SO
4
)............................................................................................................................ 0.002% max
Heavy metals (as Pb)................................................................................................................... 5 ppm max
Iron (Fe).................................................................................................................................... 5 ppm max
Calcium (Ca)............................................................................................................................ 0.005% max
Magnesium (Mg)...................................................................................................................... 0.002% max
Potassium (K)........................................................................................................................... 0.005% max
Size
Packaging
Cat. No.
Price
500 g
Poly Bottle
Ea./
47.64
Cs. 12/
437.38
2.5 kg
Poly Bottle
Ea./
106.11
Cs. 4/
371.24
SODIUM AZIDE
5% AQUEOUS SOLUTION
Formula:
NaN
3
MW:
65.01 g/mol
Boiling Pt:
~ 100°C (1013 hPa)
Sodium Azide, Solution. 5% (w/v).
N.I.S.T. traceable solution, made with purified water.
Appearance/color................................................................................................................................Clear
Shelf-life..........................................................................................................................................2 years
Sodium azide...................................................................................................................................4.9-5.1
Size
Packaging
Cat. No.
Price
500 mL
Poly Bottle
Ea./
41.61
Cs. 12/
474.38
SODIUM BICARBONATE
Sodium hydrogen carbonate
Formula:
NaHCO
3
MW:
84.01 g/mol
Melting Pt:
270°C
Density:
2.159 g/cm³ (20°C)
CAS:
144-55-8
EINECS:
205-633-8
Sodium Bicarbonate, ACS Grade.
White solid
Assay........................................................................................................................99.7 – 100.3% NaHCO
3
Insoluble matter....................................................................................................................... 0.015% max
Chloride (Cl)............................................................................................................................. 0.003% max
Phosphate (PO
4
)....................................................................................................................... 0.001% max
Sulfur compounds (as SO
4
)......................................................................................................... 0.003% max
Ammonium (NH
4
)....................................................................................................................... 5 ppm max
Heavy metals (as Pb)................................................................................................................... 5 ppm max
Iron (Fe).................................................................................................................................. 0.001% max
Calcium (Ca).............................................................................................................................. 0.02% max
Magnesium (Mg)...................................................................................................................... 0.005% max
Potassium (K)........................................................................................................................... 0.005% max
Size
Packaging
Cat. No.
Price
500 g
Poly Bottle
Ea./
33.19
Cs. 12/
281.39
2.5 kg
Poly Bottle
Ea./
62.82
Cs. 4/
219.54
12 kg
Poly Pail
Ea./
147.95
Pl. 27/
3495.39
Density:
~ 1.04 g/cm³ (20°C)
CAS:
26628-22-8
EINECS:
247-852-1
O
HO ONa
Ordering chemicals is easy
through VWR.com
Our web site gives you maximum
control over the ordering process.
With all of the available options,
you can simplify procurement by
matching the process to your
own business practices.
800.932.5000
|
vwr.com
|
VWR International
|
53
S
Sodium Bicarbonate
NaN
3
1...,61,62,63,64,65,66,67,68,69,70 72,73,74,75,76,77,78,79,80,81,...100
Powered by FlippingBook