Page 25 - BDH Chemicals Catalog 2012-2013
Basic HTML Version
ACETIC ACID, 99%
Formula:
H₃CCOOH
MW:
60.05 g/mol
Boiling Pt:
117 to 118 °C (1013 hPa)
Melting Pt:
17°C
Density:
1.05 g/cm³ (20°C)
Flash Pt:
38.5°C
Molybdenum (Mo)........................................................................................................... 10 ppt (pg/g) max.
Neodymium (Nd)............................................................................................................... 1 ppt (pg/g) max.
Nickel (Ni). ..................................................................................................................... 50 ppt (pg/g) max.
Platinum (Pt)................................................................................................................... 50 ppt (pg/g) max.
Potassium (K).................................................................................................................. 50 ppt (pg/g) max.
Praseodymium (Pr). ........................................................................................................... 1 ppt (pg/g) max.
Rhenium (Re).................................................................................................................. 10 ppt (pg/g) max.
Rhodium (Rh).................................................................................................................. 50 ppt (pg/g) max.
Rubidium (Rb)................................................................................................................. 10 ppt (pg/g) max.
Ruthenium (Ru)............................................................................................................... 50 ppt (pg/g) max.
Samarium (Sm). ................................................................................................................ 1 ppt (pg/g) max.
Scandium (Sc). ................................................................................................................ 10 ppt (pg/g) max.
Selenium (Se)...................................................................................................................... Information only
Silver (Ag)....................................................................................................................... 50 ppt (pg/g) max.
Sodium (Na).................................................................................................................. 100 ppt (pg/g) max.
Strontium (Sr).................................................................................................................. 10 ppt (pg/g) max.
Tellurium (Te).................................................................................................................... 1 ppt (pg/g) max.
Terbium (Tb). .................................................................................................................... 1 ppt (pg/g) max.
Thallium (Tl). .................................................................................................................. 10 ppt (pg/g) max.
Thorium (Th)..................................................................................................................... 1 ppt (pg/g) max.
Thulium (Tm). ................................................................................................................... 1 ppt (pg/g) max.
Tin (Sn). ......................................................................................................................... 50 ppt (pg/g) max.
Titanium (Ti)................................................................................................................... 10 ppt (pg/g) max.
Tungsten (W). ................................................................................................................. 10 ppt (pg/g) max.
Uranium (U)...................................................................................................................... 1 ppt (pg/g) max.
Vanadium (V).................................................................................................................. 10 ppt (pg/g) max.
Ytterbium (Yb). ................................................................................................................. 1 ppt (pg/g) max.
Yttrium (Y)........................................................................................................................ 1 ppt (pg/g) max.
Zinc (Zn)......................................................................................................................... 50 ppt (pg/g) max.
Zirconium (Zr). ................................................................................................................ 10 ppt (pg/g) max.
Acetic Acid, BDH ARISTAR® ULTRA
Ultra high purity acid for quantitative trace metal analysis at the parts-per-trillion (ppt) level.
Assay (CH
3
COOH, w/w)...................................................................................................................99% min.
Aluminum (Al)................................................................................................................. 50 ppt (pg/g) max.
Antimony (Sb)................................................................................................................. 50 ppt (pg/g) max.
Arsenic (As). ................................................................................................................... 50 ppt (pg/g) max.
Barium (Ba). ................................................................................................................... 10 ppt (pg/g) max.
Beryllium (Be). ................................................................................................................ 10 ppt (pg/g) max.
Bismuth (Bi).................................................................................................................... 10 ppt (pg/g) max.
Cadmium (Cd)................................................................................................................. 10 ppt (pg/g) max.
Calcium (Ca). .................................................................................................................. 50 ppt (pg/g) max.
Cerium (Ce)..................................................................................................................... 10 ppt (pg/g) max.
Cesium (Cs)..................................................................................................................... 10 ppt (pg/g) max.
Chromium (Cr)................................................................................................................. 10 ppt (pg/g) max.
Cobalt (Co)...................................................................................................................... 10 ppt (pg/g) max.
Copper (Cu)..................................................................................................................... 10 ppt (pg/g) max.
Dysprosium (Dy)................................................................................................................ 1 ppt (pg/g) max.
Erbium (Er). ...................................................................................................................... 1 ppt (pg/g) max.
Europium (Eu). .................................................................................................................. 1 ppt (pg/g) max.
Gadolinium (Gd)................................................................................................................ 1 ppt (pg/g) max.
Gallium (Ga). .................................................................................................................. 10 ppt (pg/g) max.
Germanium (Ge). ............................................................................................................ 10 ppt (pg/g) max.
Hafnium (Hf)................................................................................................................... 10 ppt (pg/g) max.
Holmium (Ho). .................................................................................................................. 1 ppt (pg/g) max.
Indium (In)........................................................................................................................ 1 ppt (pg/g) max.
Iron (Fe). ........................................................................................................................ 50 ppt (pg/g) max.
Lanthanum (La)................................................................................................................. 1 ppt (pg/g) max.
Lead (Pb)........................................................................................................................ 10 ppt (pg/g) max.
Lithium (Li). .................................................................................................................... 10 ppt (pg/g) max.
Lutetium (Lu). ................................................................................................................. 10 ppt (pg/g) max.
Magnesium (Mg)............................................................................................................. 50 ppt (pg/g) max.
Manganese (Mn)............................................................................................................. 10 ppt (pg/g) max.
CAS:
64-19-7
EINECS:
200-580-7
ADR:
8,II
Index:
607-002-00-6
UN:
2789
Size
Packaging
Cat. No.
Price
500 mL
FEP Bottle
Ea./
578.47
Safety meets style with
VWR® Protective Eyewear
Protective eyewear are designed for both men and women. Impact-
resistant, scratch-resistant polycarbonate lenses offer 99.9% UV protection.
Wraparound visitor specs fit over most prescription glasses. Eyewear with
wraparound lenses feature rubberized temple tips and integrated nosepiece
for comfort and a sleek appearance. Eyewear with adjustable temples that
lock into place also feature ultra-lightweight nylon frames and soft PVC
nosepads to prevent slipping.
Description
Cat. No.
Visitor Specs
89187-988
Eyewear with Wraparound Lens
89187-986
Eyewear with Adjustable Temples
89187-984
For more information, visit
us.vwr.com
HO
O
800.932.5000
|
vwr.com
|
VWR International
|
7
A
Acetic Acid