The NEW Acro 500 Capsule has been specifically designed for efficient, cost-effective filtration and high throughput of liquid volumes, remove particulate contamination, clarify solutions, and sterilize liquid solutions prior to further processing. The Acro 500 is uniquely housed in a clear polycarbonate housing; where easy observation is now possible to ensure peak performance by maximizing the entire effective filtration area (EFA) ideal for high turbidity, high bioburden solutions.