MWCO
| Application
|
1K-100K |
Biomolecule research, available in in a wide range of MWCO. High recovery of specific biomolecules |
Pore Size
| Application
|
30-40 μm |
Gross particulate clean up; i.e. plant and cannabis applications |
Pore Size
| Application
|
0.8-5.0 μm |
Sample cleanup, particulate removal, pre-filtration; various cell-based assays, resin screening, bead-based assays. |
0.45 μm |
Large virus filtration, i.e use in cell and gene therapy workflows. |
0.65/0.2 μm* |
Sterile filtration, ideal for cell culture & protein purification needs, ensure sterile effluent. Dual layer Supor EKV membrane |
0.1 μm |
Mycoplasma reduction and sterile filtration for cell culture media and feeds |
Pore Size
| Application
|
Depth media over 0.65/0.2 μm |
Clarification of raw cell cultures and sterile filtration of proteins and other biomolecules in one filter plate. |