Whether pursuing analytical goals in life science, pharmaceutical methods, research and development, quality control, or environmental applications, Pall’s sample prep Acrodisc® syringe filters offer fast reliable and reproducible results. Pall achieves this with higher flow rates (up to 245 ml/min) and greater sample throughput capacity (up to 150ml) reducing time and effort required to filter a wide array of sample volumes. Our HPLC, UPLC, LCMS and IC certified Acrodisc syringe filters efficiently remove particulate contamination from samples, protecting instruments and preventing costly system maintenance and premature column replacement extending column life up to 52 times.
Fields marked with * are mandatory